Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 169938317-169938542 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SAMD7
Entrez Gene ID
344658
Gene Name
sterile alpha motif domain containing 7
Gene Aliases
-
UniGene
Hs.439922
Gene Chromosome Location
Chr.3: 169911572-169939175 on Build GRCh38
Transcripts
NM_001304366.1 NM_182610.3 NR_130713.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5905565 c.1201C>T p.P401S Chr.3: 169938366-169938366 on Build GRCh38
4763502 c.1220A>T p.D407V Chr.3: 169938385-169938385 on Build GRCh38
235309 c.1314A>G p.G438G Chr.3: 169938479-169938479 on Build GRCh38
2156982 c.1220A>G p.D407G Chr.3: 169938385-169938385 on Build GRCh38
3915344 c.1275G>A p.M425I Chr.3: 169938440-169938440 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760072418 Chr.3: 169938368-169938368 on Build GRCh38
rs200235684 Chr.3: 169938486-169938486 on Build GRCh38
rs372316730 Chr.3: 169938465-169938465 on Build GRCh38
rs749020125 Chr.3: 169938345-169938345 on Build GRCh38
rs377614706 Chr.3: 169938462-169938462 on Build GRCh38
rs748060292 Chr.3: 169938505-169938505 on Build GRCh38
rs140888945 Chr.3: 169938435-169938435 on Build GRCh38
rs781151685 Chr.3: 169938356-169938359 on Build GRCh38
rs558421009 Chr.3: 169938503-169938503 on Build GRCh38
rs777663519 Chr.3: 169938458-169938458 on Build GRCh38
rs776118861 Chr.3: 169938490-169938490 on Build GRCh38
rs150124826 Chr.3: 169938476-169938476 on Build GRCh38
rs373905806 Chr.3: 169938487-169938487 on Build GRCh38
rs1022018335 Chr.3: 169938416-169938416 on Build GRCh38
rs138055938 Chr.3: 169938400-169938400 on Build GRCh38
rs776205466 Chr.3: 169938370-169938370 on Build GRCh38
rs750531970 Chr.3: 169938406-169938406 on Build GRCh38
rs1031075112 Chr.3: 169938439-169938439 on Build GRCh38
rs761106889 Chr.3: 169938376-169938376 on Build GRCh38
rs900351261 Chr.3: 169938411-169938411 on Build GRCh38
rs370549841 Chr.3: 169938481-169938481 on Build GRCh38
rs770317102 Chr.3: 169938347-169938347 on Build GRCh38
rs540015841 Chr.3: 169938483-169938483 on Build GRCh38
rs765178959 Chr.3: 169938405-169938405 on Build GRCh38
rs764240406 Chr.3: 169938369-169938369 on Build GRCh38
rs781106693 Chr.3: 169938422-169938422 on Build GRCh38
rs755881999 Chr.3: 169938440-169938440 on Build GRCh38
rs772730222 Chr.3: 169938344-169938344 on Build GRCh38
rs777073595 Chr.3: 169938510-169938510 on Build GRCh38
rs761300912 Chr.3: 169938492-169938492 on Build GRCh38
rs1051769509 Chr.3: 169938485-169938485 on Build GRCh38
rs764565263 Chr.3: 169938393-169938393 on Build GRCh38
rs999174866 Chr.3: 169938436-169938436 on Build GRCh38
rs867914820 Chr.3: 169938382-169938382 on Build GRCh38
rs754844902 Chr.3: 169938414-169938414 on Build GRCh38
rs374151168 Chr.3: 169938362-169938362 on Build GRCh38
rs753971854 Chr.3: 169938399-169938399 on Build GRCh38

View Full Product Details