Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 46231883-46232110 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPARA
Entrez Gene ID
5465
Gene Name
peroxisome proliferator activated receptor alpha
Gene Aliases
NR1C1, PPAR, PPARalpha, hPPAR
UniGene
Hs.103110
Gene Chromosome Location
Chr.22: 46150534-46243756 on Build GRCh38
Transcripts
NM_001001928.2 NM_005036.4 XM_005261655.3 XM_005261656.3 XM_006724269.3 XM_006724270.3 XM_011530239.2 XM_011530240.2 XM_011530241.2 XM_011530242.2 XM_011530243.2 XM_011530244.2 XM_011530245.2 XM_017028839.1 XM_017028840.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5859577 c.812G>A p.R271H Chr.22: 46231892-46231892 on Build GRCh38
4104816 c.934G>A p.G312R Chr.22: 46232014-46232014 on Build GRCh38
3063391 c.933C>T p.Y311Y Chr.22: 46232013-46232013 on Build GRCh38
479081 c.984C>T p.D328D Chr.22: 46232064-46232064 on Build GRCh38
3555410 c.1030C>T p.L344L Chr.22: 46232110-46232110 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747311682 Chr.22: 46231941-46231941 on Build GRCh38
rs201836521 Chr.22: 46232055-46232055 on Build GRCh38
rs774720320 Chr.22: 46231971-46231971 on Build GRCh38
rs755317696 Chr.22: 46232025-46232025 on Build GRCh38
rs373877541 Chr.22: 46232082-46232082 on Build GRCh38
rs563662966 Chr.22: 46232079-46232079 on Build GRCh38
rs769733697 Chr.22: 46231951-46231951 on Build GRCh38
rs373080217 Chr.22: 46231944-46231944 on Build GRCh38
rs764002662 Chr.22: 46231989-46231989 on Build GRCh38
rs778532987 Chr.22: 46231917-46231917 on Build GRCh38
rs764645826 Chr.22: 46231992-46231992 on Build GRCh38
rs751238606 Chr.22: 46232023-46232023 on Build GRCh38
rs757913269 Chr.22: 46231999-46231999 on Build GRCh38
rs369962398 Chr.22: 46232046-46232046 on Build GRCh38
rs1800242 Chr.22: 46231990-46231990 on Build GRCh38
rs937915400 Chr.22: 46232035-46232035 on Build GRCh38
rs750003622 Chr.22: 46231993-46231993 on Build GRCh38
rs199894775 Chr.22: 46231950-46231950 on Build GRCh38
rs773411072 Chr.22: 46231955-46231955 on Build GRCh38
rs777078159 Chr.22: 46231947-46231947 on Build GRCh38
rs756672001 Chr.22: 46231916-46231916 on Build GRCh38
rs370814026 Chr.22: 46232078-46232078 on Build GRCh38
rs779611146 Chr.22: 46232019-46232019 on Build GRCh38
rs913307394 Chr.22: 46231967-46231967 on Build GRCh38
rs763073521 Chr.22: 46231956-46231956 on Build GRCh38
rs781580066 Chr.22: 46232038-46232038 on Build GRCh38
rs925990203 Chr.22: 46232013-46232013 on Build GRCh38
rs917522785 Chr.22: 46232064-46232064 on Build GRCh38
rs376285178 Chr.22: 46232057-46232057 on Build GRCh38
rs544843326 Chr.22: 46231935-46231935 on Build GRCh38
rs567297876 Chr.22: 46231934-46231934 on Build GRCh38
rs753857353 Chr.22: 46231912-46231912 on Build GRCh38
rs4253773 Chr.22: 46231983-46231983 on Build GRCh38
rs376174372 Chr.22: 46231932-46231932 on Build GRCh38
rs145265093 Chr.22: 46232073-46232073 on Build GRCh38
rs749323438 Chr.22: 46232065-46232065 on Build GRCh38
rs763603755 Chr.22: 46232034-46232034 on Build GRCh38
rs575149167 Chr.22: 46231965-46231965 on Build GRCh38

View Full Product Details