Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 86681699-86681944 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LDB3
Entrez Gene ID
11155
Gene Name
LIM domain binding 3
Gene Aliases
CMD1C, CMH24, CMPD3, CYPHER, LDB3Z1, LDB3Z4, LVNC3, MFM4, ORACLE, PDLIM6, ZASP
UniGene
Hs.657271
Gene Chromosome Location
Chr.10: 86664785-86736072 on Build GRCh38
Transcripts
NM_001080114.1 NM_001080115.1 NM_001080116.1 NM_001171610.1 NM_001171611.1 NM_007078.2 XM_005269464.4 XM_005269466.4 XM_005269468.4 XM_011539184.2 XM_011539185.2 XM_011539186.2 XM_011539187.2 XM_011539188.2 XM_011539189.1 XM_011539190.1 XM_011539191.2 XM_011539193.1 XM_011539194.1 XM_011539195.2 XM_017015606.1 XM_017015608.1 XM_017015609.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6130304 c.681C>A p.L227L Chr.10: 86681795-86681795 on Build GRCh38
6218619 c.663G>C p.K221N Chr.10: 86681777-86681777 on Build GRCh38
4842168 c.627G>C p.E209D Chr.10: 86681741-86681741 on Build GRCh38
1492354 c.610G>A p.A204T Chr.10: 86681724-86681724 on Build GRCh38
1492355 c.610G>A p.A204T Chr.10: 86681724-86681724 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770926992 Chr.10: 86681907-86681907 on Build GRCh38
rs759858111 Chr.10: 86681726-86681726 on Build GRCh38
rs767919085 Chr.10: 86681728-86681728 on Build GRCh38
rs529449847 Chr.10: 86681891-86681891 on Build GRCh38
rs754319407 Chr.10: 86681920-86681920 on Build GRCh38
rs765199175 Chr.10: 86681760-86681760 on Build GRCh38
rs45563234 Chr.10: 86681813-86681813 on Build GRCh38
rs761383536 Chr.10: 86681834-86681834 on Build GRCh38
rs1021921417 Chr.10: 86681864-86681864 on Build GRCh38
rs763616272 Chr.10: 86681733-86681733 on Build GRCh38
rs727503124 Chr.10: 86681812-86681812 on Build GRCh38
rs957930268 Chr.10: 86681759-86681759 on Build GRCh38
rs903272791 Chr.10: 86681875-86681875 on Build GRCh38
rs45548935 Chr.10: 86681885-86681885 on Build GRCh38
rs776235220 Chr.10: 86681833-86681833 on Build GRCh38
rs774976112 Chr.10: 86681724-86681724 on Build GRCh38
rs778089838 Chr.10: 86681783-86681783 on Build GRCh38
rs727503123 Chr.10: 86681769-86681769 on Build GRCh38
rs778367138 Chr.10: 86681839-86681839 on Build GRCh38
rs201818569 Chr.10: 86681915-86681915 on Build GRCh38
rs121908337 Chr.10: 86681731-86681731 on Build GRCh38
rs940818176 Chr.10: 86681738-86681738 on Build GRCh38
rs139922045 Chr.10: 86681778-86681778 on Build GRCh38
rs749425278 Chr.10: 86681789-86681789 on Build GRCh38
rs953482767 Chr.10: 86681912-86681912 on Build GRCh38
rs528600962 Chr.10: 86681863-86681863 on Build GRCh38
rs753369970 Chr.10: 86681739-86681739 on Build GRCh38
rs551288539 Chr.10: 86681892-86681892 on Build GRCh38
rs147300824 Chr.10: 86681749-86681749 on Build GRCh38
rs201538257 Chr.10: 86681722-86681722 on Build GRCh38
rs895933043 Chr.10: 86681825-86681825 on Build GRCh38
rs967244938 Chr.10: 86681873-86681873 on Build GRCh38
rs375306400 Chr.10: 86681782-86681782 on Build GRCh38
rs768354453 Chr.10: 86681722-86681732 on Build GRCh38
rs779630176 Chr.10: 86681804-86681804 on Build GRCh38
rs530979771 Chr.10: 86681770-86681770 on Build GRCh38
rs762955577 Chr.10: 86681852-86681852 on Build GRCh38
rs1048074821 Chr.10: 86681793-86681793 on Build GRCh38
rs775061423 Chr.10: 86681729-86681729 on Build GRCh38
rs749848991 Chr.10: 86681846-86681846 on Build GRCh38
rs146668620 Chr.10: 86681843-86681843 on Build GRCh38
rs1020443599 Chr.10: 86681856-86681856 on Build GRCh38
rs45531131 Chr.10: 86681723-86681723 on Build GRCh38
rs888143005 Chr.10: 86681794-86681794 on Build GRCh38
rs771099131 Chr.10: 86681790-86681790 on Build GRCh38
rs547089159 Chr.10: 86681879-86681879 on Build GRCh38
rs1036079839 Chr.10: 86681742-86681742 on Build GRCh38
rs375094467 Chr.10: 86681818-86681818 on Build GRCh38
rs1031129987 Chr.10: 86681884-86681884 on Build GRCh38
rs750360125 Chr.10: 86681768-86681768 on Build GRCh38
rs760901326 Chr.10: 86681814-86681814 on Build GRCh38

View Full Product Details