Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 55912800-55913055 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NLRP13
Entrez Gene ID
126204
Gene Name
NLR family pyrin domain containing 13
Gene Aliases
CLR19.7, NALP13, NOD14, PAN13
UniGene
Hs.446924
Gene Chromosome Location
Chr.19: 55891692-55932336 on Build GRCh38
Transcripts
NM_001321057.1 NM_176810.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4599076 c.847G>A p.E283K Chr.19: 55912970-55912970 on Build GRCh38
1001730 c.983A>G p.D328G Chr.19: 55912834-55912834 on Build GRCh38
7152159 c.890A>G p.E297G Chr.19: 55912927-55912927 on Build GRCh38
3673023 c.971A>C p.E324A Chr.19: 55912846-55912846 on Build GRCh38
6053136 c.784T>G p.F262V Chr.19: 55913033-55913033 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs116162869 Chr.19: 55912827-55912827 on Build GRCh38
rs754187987 Chr.19: 55912909-55912909 on Build GRCh38
rs774291227 Chr.19: 55913006-55913006 on Build GRCh38
rs779639451 Chr.19: 55912901-55912901 on Build GRCh38
rs142186945 Chr.19: 55912944-55912944 on Build GRCh38
rs907936187 Chr.19: 55912908-55912908 on Build GRCh38
rs778215504 Chr.19: 55912980-55912980 on Build GRCh38
rs868746137 Chr.19: 55912910-55912910 on Build GRCh38
rs1056760924 Chr.19: 55913004-55913004 on Build GRCh38
rs866889624 Chr.19: 55912874-55912874 on Build GRCh38
rs376929098 Chr.19: 55912935-55912935 on Build GRCh38
rs147819247 Chr.19: 55912957-55912957 on Build GRCh38
rs758951506 Chr.19: 55912975-55912975 on Build GRCh38
rs753148509 Chr.19: 55912972-55912972 on Build GRCh38
rs774939659 Chr.19: 55912918-55912918 on Build GRCh38
rs147787759 Chr.19: 55912959-55912959 on Build GRCh38
rs373711481 Chr.19: 55913017-55913017 on Build GRCh38
rs753708026 Chr.19: 55912946-55912946 on Build GRCh38
rs774920200 Chr.19: 55912887-55912889 on Build GRCh38
rs765838590 Chr.19: 55913024-55913024 on Build GRCh38
rs766683816 Chr.19: 55912968-55912968 on Build GRCh38
rs746112690 Chr.19: 55912994-55912994 on Build GRCh38
rs781432326 Chr.19: 55912992-55912992 on Build GRCh38
rs761364584 Chr.19: 55912857-55912857 on Build GRCh38
rs200626553 Chr.19: 55912823-55912823 on Build GRCh38
rs200051453 Chr.19: 55912853-55912853 on Build GRCh38
rs755794031 Chr.19: 55912866-55912866 on Build GRCh38
rs767074253 Chr.19: 55912860-55912860 on Build GRCh38
rs188240771 Chr.19: 55912947-55912947 on Build GRCh38
rs747136573 Chr.19: 55912936-55912936 on Build GRCh38
rs753529486 Chr.19: 55912967-55912967 on Build GRCh38
rs776218787 Chr.19: 55913026-55913026 on Build GRCh38
rs546950202 Chr.19: 55912922-55912922 on Build GRCh38
rs761852882 Chr.19: 55912949-55912949 on Build GRCh38
rs772032663 Chr.19: 55913013-55913013 on Build GRCh38
rs761765048 Chr.19: 55913011-55913011 on Build GRCh38
rs768510676 Chr.19: 55912996-55912996 on Build GRCh38
rs777925796 Chr.19: 55912933-55912933 on Build GRCh38
rs554222717 Chr.19: 55912982-55912982 on Build GRCh38
rs764097069 Chr.19: 55912852-55912852 on Build GRCh38
rs766031545 Chr.19: 55912960-55912960 on Build GRCh38
rs371874380 Chr.19: 55912864-55912864 on Build GRCh38
rs764886195 Chr.19: 55912830-55912830 on Build GRCh38
rs149164646 Chr.19: 55912939-55912939 on Build GRCh38
rs137923694 Chr.19: 55913029-55913029 on Build GRCh38
rs566645087 Chr.19: 55912932-55912932 on Build GRCh38
rs755385447 Chr.19: 55912915-55912915 on Build GRCh38
rs779272690 Chr.19: 55912971-55912971 on Build GRCh38
rs775363998 Chr.19: 55912838-55912838 on Build GRCh38
rs145223514 Chr.19: 55912842-55912842 on Build GRCh38
rs371198255 Chr.19: 55912990-55912990 on Build GRCh38
rs760627249 Chr.19: 55912929-55912929 on Build GRCh38
rs61738368 Chr.19: 55913016-55913016 on Build GRCh38

View Full Product Details