Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 8158462-8158717 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ICA1
Entrez Gene ID
3382
Gene Name
islet cell autoantigen 1
Gene Aliases
ICA69, ICAp69
UniGene
Hs.487561
Gene Chromosome Location
Chr.7: 8113184-8262687 on Build GRCh38
Transcripts
NM_001136020.2 NM_001276478.1 NM_004968.3 NM_022307.2 XM_005249735.3 XM_011515345.1 XM_011515346.1 XM_011515347.2 XM_011515348.1 XM_011515349.1 XM_011515350.1 XM_011515351.1 XM_011515352.1 XM_011515353.2 XM_011515354.1 XM_011515355.2 XM_011515356.2 XM_011515357.2 XM_017012116.1 XM_017012117.1 XM_017012118.1 XM_017012119.1 XM_017012120.1 XM_017012121.1 XM_017012122.1 XM_017012123.1 XM_017012124.1 XM_017012125.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3640924 c.676C>G p.L226V Chr.7: 8158556-8158556 on Build GRCh38
7126747 c.653T>A p.L218H Chr.7: 8158579-8158579 on Build GRCh38
5495040 c.663G>C p.A221A Chr.7: 8158569-8158569 on Build GRCh38
5017332 c.612_613insA p.F205fs*2 Chr.7: 8158619-8158620 on Build GRCh38
1452273 c.611_612insA p.N204fs*3 Chr.7: 8158620-8158621 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769481318 Chr.7: 8158607-8158607 on Build GRCh38
rs140851537 Chr.7: 8158617-8158617 on Build GRCh38
rs186186283 Chr.7: 8158680-8158680 on Build GRCh38
rs1032440988 Chr.7: 8158544-8158544 on Build GRCh38
rs756843206 Chr.7: 8158670-8158670 on Build GRCh38
rs777602698 Chr.7: 8158656-8158656 on Build GRCh38
rs747841314 Chr.7: 8158621-8158621 on Build GRCh38
rs769979445 Chr.7: 8158492-8158492 on Build GRCh38
rs750551548 Chr.7: 8158522-8158522 on Build GRCh38
rs775412759 Chr.7: 8158610-8158610 on Build GRCh38
rs751066360 Chr.7: 8158666-8158666 on Build GRCh38
rs375940094 Chr.7: 8158495-8158495 on Build GRCh38
rs781302996 Chr.7: 8158603-8158603 on Build GRCh38
rs772473952 Chr.7: 8158614-8158614 on Build GRCh38
rs753158452 Chr.7: 8158644-8158644 on Build GRCh38
rs374891501 Chr.7: 8158566-8158566 on Build GRCh38
rs754151046 Chr.7: 8158533-8158533 on Build GRCh38
rs773234246 Chr.7: 8158683-8158683 on Build GRCh38
rs761918023 Chr.7: 8158613-8158613 on Build GRCh38
rs7798010 Chr.7: 8158596-8158596 on Build GRCh38
rs766899705 Chr.7: 8158526-8158526 on Build GRCh38
rs778755992 Chr.7: 8158581-8158581 on Build GRCh38
rs755307933 Chr.7: 8158554-8158554 on Build GRCh38
rs772022982 Chr.7: 8158620-8158621 on Build GRCh38
rs7802005 Chr.7: 8158650-8158650 on Build GRCh38
rs770953752 Chr.7: 8158601-8158601 on Build GRCh38
rs138064847 Chr.7: 8158583-8158583 on Build GRCh38
rs779682791 Chr.7: 8158686-8158686 on Build GRCh38
rs754301457 Chr.7: 8158634-8158634 on Build GRCh38
rs760763922 Chr.7: 8158625-8158625 on Build GRCh38
rs952572650 Chr.7: 8158645-8158645 on Build GRCh38
rs879048330 Chr.7: 8158535-8158535 on Build GRCh38
rs766811711 Chr.7: 8158631-8158631 on Build GRCh38
rs765509427 Chr.7: 8158642-8158642 on Build GRCh38
rs370722122 Chr.7: 8158504-8158504 on Build GRCh38
rs745618958 Chr.7: 8158604-8158604 on Build GRCh38
rs960213582 Chr.7: 8158510-8158510 on Build GRCh38
rs752291214 Chr.7: 8158569-8158569 on Build GRCh38
rs17847180 Chr.7: 8158556-8158556 on Build GRCh38
rs77882114 Chr.7: 8158629-8158629 on Build GRCh38
rs372639629 Chr.7: 8158523-8158523 on Build GRCh38
rs758131535 Chr.7: 8158572-8158572 on Build GRCh38
rs368526579 Chr.7: 8158497-8158497 on Build GRCh38
rs756126096 Chr.7: 8158683-8158683 on Build GRCh38
rs760153189 Chr.7: 8158637-8158637 on Build GRCh38
rs755603457 Chr.7: 8158493-8158493 on Build GRCh38
rs942573276 Chr.7: 8158685-8158685 on Build GRCh38
rs978771945 Chr.7: 8158571-8158571 on Build GRCh38
rs781209679 Chr.7: 8158675-8158675 on Build GRCh38
rs1025651513 Chr.7: 8158636-8158636 on Build GRCh38

View Full Product Details