Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 152990952-152991199 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PNMA5
Entrez Gene ID
114824
Gene Name
paraneoplastic Ma antigen family member 5
Gene Aliases
-
UniGene
Hs.573567
Gene Chromosome Location
Chr.X: 152988824-152994458 on Build GRCh38
Transcripts
NM_001103150.1 NM_001103151.1 NM_001184924.1 NM_052926.2 XM_017029252.1 XM_017029253.1

SNP Detail

SNP ID Chromosome Location
rs782461300 Chr.X: 152991166-152991166 on Build GRCh38
rs781829344 Chr.X: 152991086-152991086 on Build GRCh38
rs142107812 Chr.X: 152991171-152991171 on Build GRCh38
rs782622035 Chr.X: 152991094-152991094 on Build GRCh38
rs368937649 Chr.X: 152991026-152991026 on Build GRCh38
rs782085755 Chr.X: 152991180-152991180 on Build GRCh38
rs199625105 Chr.X: 152991126-152991126 on Build GRCh38
rs782164970 Chr.X: 152991023-152991023 on Build GRCh38
rs782287773 Chr.X: 152991111-152991111 on Build GRCh38
rs782176856 Chr.X: 152991141-152991141 on Build GRCh38
rs782386047 Chr.X: 152991110-152991110 on Build GRCh38
rs782698342 Chr.X: 152991170-152991170 on Build GRCh38
rs1054102157 Chr.X: 152991007-152991007 on Build GRCh38
rs782239283 Chr.X: 152991033-152991033 on Build GRCh38
rs782620683 Chr.X: 152991078-152991078 on Build GRCh38
rs782072627 Chr.X: 152991168-152991168 on Build GRCh38
rs781868637 Chr.X: 152991157-152991157 on Build GRCh38
rs782437367 Chr.X: 152991061-152991061 on Build GRCh38
rs180860237 Chr.X: 152990993-152990993 on Build GRCh38
rs782438353 Chr.X: 152991158-152991158 on Build GRCh38
rs782533954 Chr.X: 152991087-152991087 on Build GRCh38
rs376216037 Chr.X: 152990986-152990986 on Build GRCh38
rs782584813 Chr.X: 152991134-152991134 on Build GRCh38
rs781950043 Chr.X: 152991150-152991150 on Build GRCh38
rs781858850 Chr.X: 152991017-152991017 on Build GRCh38
rs782716646 Chr.X: 152991003-152991003 on Build GRCh38
rs781814267 Chr.X: 152991037-152991037 on Build GRCh38
rs148898774 Chr.X: 152991149-152991149 on Build GRCh38
rs782094822 Chr.X: 152990999-152990999 on Build GRCh38

View Full Product Details