Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 132487843-132488096 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IRF1
Entrez Gene ID
3659
Gene Name
interferon regulatory factor 1
Gene Aliases
IRF-1, MAR
UniGene
Hs.436061
Gene Chromosome Location
Chr.5: 132481609-132490789 on Build GRCh38
Transcripts
NM_002198.2 XM_011543378.2 XM_011543379.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6356533 c.187+1G>A p.? Chr.5: 132487925-132487925 on Build GRCh38
4408228 c.187G>A p.G63S Chr.5: 132487926-132487926 on Build GRCh38
5356443 c.175G>A p.A59T Chr.5: 132487938-132487938 on Build GRCh38
4832913 c.96G>C p.M32I Chr.5: 132488017-132488017 on Build GRCh38
3850047 c.129G>A p.K43K Chr.5: 132487984-132487984 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs545850001 Chr.5: 132487981-132487981 on Build GRCh38
rs772847790 Chr.5: 132488066-132488066 on Build GRCh38
rs778809271 Chr.5: 132488042-132488042 on Build GRCh38
rs1038764450 Chr.5: 132488002-132488002 on Build GRCh38
rs767077054 Chr.5: 132487897-132487897 on Build GRCh38
rs749465418 Chr.5: 132488031-132488031 on Build GRCh38
rs768572080 Chr.5: 132488036-132488036 on Build GRCh38
rs760094979 Chr.5: 132488071-132488071 on Build GRCh38
rs770420063 Chr.5: 132488075-132488075 on Build GRCh38
rs375288471 Chr.5: 132488058-132488058 on Build GRCh38
rs754850426 Chr.5: 132487890-132487890 on Build GRCh38
rs199614102 Chr.5: 132488044-132488044 on Build GRCh38
rs758703389 Chr.5: 132487887-132487887 on Build GRCh38
rs764376275 Chr.5: 132487894-132487894 on Build GRCh38
rs781296212 Chr.5: 132487935-132487935 on Build GRCh38
rs776153392 Chr.5: 132488076-132488076 on Build GRCh38
rs1055525400 Chr.5: 132487999-132487999 on Build GRCh38
rs758776198 Chr.5: 132487889-132487889 on Build GRCh38
rs756424796 Chr.5: 132487960-132487960 on Build GRCh38
rs117282832 Chr.5: 132488011-132488011 on Build GRCh38
rs780125312 Chr.5: 132487998-132487998 on Build GRCh38
rs938557866 Chr.5: 132487867-132487867 on Build GRCh38
rs750437158 Chr.5: 132487946-132487946 on Build GRCh38
rs1011241942 Chr.5: 132488040-132488040 on Build GRCh38
rs757714691 Chr.5: 132487909-132487909 on Build GRCh38
rs56399643 Chr.5: 132487866-132487866 on Build GRCh38

View Full Product Details