Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 67563168-67563441 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAP2K5
Entrez Gene ID
5607
Gene Name
mitogen-activated protein kinase kinase 5
Gene Aliases
HsT17454, MAPKK5, MEK5, PRKMK5
UniGene
Hs.114198
Gene Chromosome Location
Chr.15: 67542606-67807117 on Build GRCh38
Transcripts
NM_001206804.1 NM_002757.3 NM_145160.2 XM_011521787.2 XM_011521788.2 XM_017022414.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
234271 c.205C>T p.R69* Chr.15: 67563303-67563303 on Build GRCh38
6374394 c.244C>A p.L82M Chr.15: 67563342-67563342 on Build GRCh38
6001745 c.194A>G p.E65G Chr.15: 67563292-67563292 on Build GRCh38
964291 c.185-1G>T p.? Chr.15: 67563282-67563282 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs977455924 Chr.15: 67563372-67563372 on Build GRCh38
rs150402318 Chr.15: 67563326-67563326 on Build GRCh38
rs369050562 Chr.15: 67563393-67563393 on Build GRCh38
rs748906365 Chr.15: 67563365-67563365 on Build GRCh38
rs200944611 Chr.15: 67563262-67563262 on Build GRCh38
rs770404451 Chr.15: 67563251-67563251 on Build GRCh38
rs201621976 Chr.15: 67563257-67563257 on Build GRCh38
rs188242878 Chr.15: 67563366-67563366 on Build GRCh38
rs769137126 Chr.15: 67563389-67563389 on Build GRCh38
rs777310126 Chr.15: 67563215-67563215 on Build GRCh38
rs372495824 Chr.15: 67563338-67563338 on Build GRCh38
rs1056034239 Chr.15: 67563342-67563342 on Build GRCh38
rs765565881 Chr.15: 67563286-67563286 on Build GRCh38
rs199674502 Chr.15: 67563335-67563335 on Build GRCh38
rs769429769 Chr.15: 67563232-67563232 on Build GRCh38
rs758602136 Chr.15: 67563307-67563307 on Build GRCh38
rs199953786 Chr.15: 67563272-67563272 on Build GRCh38
rs772734485 Chr.15: 67563237-67563237 on Build GRCh38
rs55744419 Chr.15: 67563347-67563347 on Build GRCh38
rs1036900410 Chr.15: 67563276-67563276 on Build GRCh38
rs374165114 Chr.15: 67563360-67563360 on Build GRCh38
rs922701284 Chr.15: 67563208-67563208 on Build GRCh38
rs375398195 Chr.15: 67563392-67563392 on Build GRCh38
rs754169921 Chr.15: 67563271-67563271 on Build GRCh38
rs777607027 Chr.15: 67563374-67563375 on Build GRCh38
rs761180901 Chr.15: 67563369-67563369 on Build GRCh38
rs367973577 Chr.15: 67563337-67563337 on Build GRCh38
rs886995566 Chr.15: 67563269-67563269 on Build GRCh38
rs751579877 Chr.15: 67563336-67563336 on Build GRCh38
rs762506312 Chr.15: 67563250-67563250 on Build GRCh38
rs773798356 Chr.15: 67563368-67563368 on Build GRCh38
rs748041362 Chr.15: 67563339-67563339 on Build GRCh38
rs762347555 Chr.15: 67563245-67563245 on Build GRCh38
rs750663318 Chr.15: 67563296-67563296 on Build GRCh38
rs374139864 Chr.15: 67563211-67563211 on Build GRCh38
rs1038178212 Chr.15: 67563220-67563220 on Build GRCh38
rs960256266 Chr.15: 67563406-67563406 on Build GRCh38

View Full Product Details