Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 110403856-110404107 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LAMTOR5
Entrez Gene ID
10542
Gene Name
late endosomal/lysosomal adaptor, MAPK and MTOR activator 5
Gene Aliases
HBXIP, XIP
UniGene
Hs.439815
Gene Chromosome Location
Chr.1: 110401253-110407924 on Build GRCh38
Transcripts
NM_006402.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
292017 c.386T>A p.V129D Chr.1: 110403994-110403994 on Build GRCh38
4004681 c.361G>C p.D121H Chr.1: 110404019-110404019 on Build GRCh38
893671 c.461G>A p.G154E Chr.1: 110403919-110403919 on Build GRCh38
5992909 c.456T>C p.D152D Chr.1: 110403924-110403924 on Build GRCh38
3934003 c.345C>T p.C115C Chr.1: 110404035-110404035 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768736997 Chr.1: 110404080-110404080 on Build GRCh38
rs761857902 Chr.1: 110404020-110404020 on Build GRCh38
rs553882943 Chr.1: 110404051-110404051 on Build GRCh38
rs111871923 Chr.1: 110404014-110404014 on Build GRCh38
rs542357923 Chr.1: 110403939-110403939 on Build GRCh38
rs763944185 Chr.1: 110403914-110403914 on Build GRCh38
rs775788107 Chr.1: 110404002-110404002 on Build GRCh38
rs778243046 Chr.1: 110404045-110404045 on Build GRCh38
rs753545435 Chr.1: 110404034-110404034 on Build GRCh38
rs779703357 Chr.1: 110404045-110404046 on Build GRCh38
rs1034621093 Chr.1: 110403984-110403984 on Build GRCh38
rs776262301 Chr.1: 110404073-110404073 on Build GRCh38
rs1802101 Chr.1: 110403957-110403957 on Build GRCh38
rs368451165 Chr.1: 110403932-110403932 on Build GRCh38
rs746873837 Chr.1: 110404074-110404075 on Build GRCh38
rs766029896 Chr.1: 110404033-110404033 on Build GRCh38
rs149410124 Chr.1: 110404032-110404032 on Build GRCh38
rs371814505 Chr.1: 110403935-110403935 on Build GRCh38
rs776825739 Chr.1: 110403988-110403988 on Build GRCh38
rs758729220 Chr.1: 110403941-110403941 on Build GRCh38
rs754579531 Chr.1: 110404039-110404039 on Build GRCh38
rs770173012 Chr.1: 110404001-110404001 on Build GRCh38
rs201619706 Chr.1: 110403958-110403958 on Build GRCh38
rs749563407 Chr.1: 110404076-110404076 on Build GRCh38
rs759882046 Chr.1: 110403886-110403886 on Build GRCh38
rs781750841 Chr.1: 110404049-110404049 on Build GRCh38
rs769938983 Chr.1: 110404058-110404058 on Build GRCh38
rs762796830 Chr.1: 110403901-110403901 on Build GRCh38
rs572734967 Chr.1: 110404040-110404040 on Build GRCh38
rs907149350 Chr.1: 110403972-110403972 on Build GRCh38
rs757136030 Chr.1: 110403925-110403925 on Build GRCh38
rs771314460 Chr.1: 110403967-110403967 on Build GRCh38
rs561079795 Chr.1: 110403998-110403998 on Build GRCh38
rs751457710 Chr.1: 110403918-110403918 on Build GRCh38
rs780268990 Chr.1: 110404063-110404063 on Build GRCh38
rs777984013 Chr.1: 110403949-110403949 on Build GRCh38
rs767308127 Chr.1: 110404024-110404024 on Build GRCh38
rs773985055 Chr.1: 110404015-110404015 on Build GRCh38
rs749912064 Chr.1: 110403928-110403928 on Build GRCh38
rs758424408 Chr.1: 110404041-110404041 on Build GRCh38
rs1002780237 Chr.1: 110403974-110403974 on Build GRCh38
rs775448726 Chr.1: 110403893-110403893 on Build GRCh38
rs372177849 Chr.1: 110403926-110403926 on Build GRCh38
rs11541367 Chr.1: 110403962-110403962 on Build GRCh38
rs200442714 Chr.1: 110403930-110403930 on Build GRCh38
rs749966907 Chr.1: 110404031-110404031 on Build GRCh38
rs762857071 Chr.1: 110404013-110404013 on Build GRCh38
rs368528095 Chr.1: 110403891-110403891 on Build GRCh38

View Full Product Details