Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 165544267-165544536 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC400794
Entrez Gene ID
400794
Gene Name
uncharacterized LOC400794
Gene Aliases
-
UniGene
Hs.374847
Gene Chromosome Location
Chr.1: 165476842-165582155 on Build GRCh38
Transcripts
NR_026744.2 NM_001005214.3

Gene Symbol
LRRC52
Entrez Gene ID
440699
Gene Name
leucine rich repeat containing 52
Gene Aliases
-
UniGene
Hs.664975
Gene Chromosome Location
Chr.1: 165544241-165563948 on Build GRCh38
Transcripts
NR_026744.2 NM_001005214.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3710390 c.217C>G p.L73V Chr.1: 165544513-165544513 on Build GRCh38
4626885 c.153A>G p.I51M Chr.1: 165544449-165544449 on Build GRCh38
1335907 c.150C>T p.D50D Chr.1: 165544446-165544446 on Build GRCh38
6703974 c.121G>T p.G41W Chr.1: 165544417-165544417 on Build GRCh38
4530331 c.168G>A p.R56R Chr.1: 165544464-165544464 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747363519 Chr.1: 165544320-165544320 on Build GRCh38
rs372760189 Chr.1: 165544500-165544500 on Build GRCh38
rs148736377 Chr.1: 165544473-165544473 on Build GRCh38
rs575834588 Chr.1: 165544453-165544453 on Build GRCh38
rs760253181 Chr.1: 165544460-165544460 on Build GRCh38
rs755617173 Chr.1: 165544295-165544295 on Build GRCh38
rs771000900 Chr.1: 165544370-165544370 on Build GRCh38
rs767831037 Chr.1: 165544364-165544364 on Build GRCh38
rs763532309 Chr.1: 165544402-165544402 on Build GRCh38
rs781473289 Chr.1: 165544417-165544417 on Build GRCh38
rs376485734 Chr.1: 165544423-165544423 on Build GRCh38
rs758918591 Chr.1: 165544307-165544307 on Build GRCh38
rs762930278 Chr.1: 165544360-165544360 on Build GRCh38
rs780560522 Chr.1: 165544408-165544408 on Build GRCh38
rs373800156 Chr.1: 165544434-165544434 on Build GRCh38
rs780329598 Chr.1: 165544317-165544317 on Build GRCh38
rs770945660 Chr.1: 165544437-165544437 on Build GRCh38
rs137987001 Chr.1: 165544326-165544326 on Build GRCh38
rs980730690 Chr.1: 165544445-165544445 on Build GRCh38
rs901622277 Chr.1: 165544287-165544287 on Build GRCh38
rs868335002 Chr.1: 165544461-165544461 on Build GRCh38
rs755340784 Chr.1: 165544416-165544416 on Build GRCh38
rs200101224 Chr.1: 165544404-165544404 on Build GRCh38
rs760214975 Chr.1: 165544401-165544401 on Build GRCh38
rs758861323 Chr.1: 165544406-165544406 on Build GRCh38
rs376324521 Chr.1: 165544505-165544505 on Build GRCh38
rs752356470 Chr.1: 165544394-165544394 on Build GRCh38
rs139452874 Chr.1: 165544440-165544440 on Build GRCh38
rs889342994 Chr.1: 165544363-165544363 on Build GRCh38
rs770158593 Chr.1: 165544430-165544430 on Build GRCh38
rs752226406 Chr.1: 165544294-165544294 on Build GRCh38
rs760771607 Chr.1: 165544432-165544432 on Build GRCh38
rs374191095 Chr.1: 165544303-165544303 on Build GRCh38
rs753113632 Chr.1: 165544489-165544489 on Build GRCh38
rs751928456 Chr.1: 165544415-165544415 on Build GRCh38
rs150814230 Chr.1: 165544479-165544479 on Build GRCh38
rs977665553 Chr.1: 165544485-165544485 on Build GRCh38
rs767109659 Chr.1: 165544291-165544291 on Build GRCh38
rs142543958 Chr.1: 165544463-165544463 on Build GRCh38
rs749360058 Chr.1: 165544506-165544506 on Build GRCh38
rs748377306 Chr.1: 165544419-165544419 on Build GRCh38
rs958112685 Chr.1: 165544358-165544358 on Build GRCh38
rs759275355 Chr.1: 165544383-165544383 on Build GRCh38
rs748229362 Chr.1: 165544333-165544333 on Build GRCh38
rs777935960 Chr.1: 165544431-165544431 on Build GRCh38
rs367686135 Chr.1: 165544462-165544462 on Build GRCh38
rs970815137 Chr.1: 165544305-165544305 on Build GRCh38
rs141692696 Chr.1: 165544353-165544353 on Build GRCh38
rs775599204 Chr.1: 165544385-165544385 on Build GRCh38
rs779384837 Chr.1: 165544301-165544301 on Build GRCh38
rs773269360 Chr.1: 165544359-165544359 on Build GRCh38
rs932952874 Chr.1: 165544315-165544315 on Build GRCh38
rs763795745 Chr.1: 165544433-165544433 on Build GRCh38
rs897976995 Chr.1: 165544396-165544396 on Build GRCh38
rs766842411 Chr.1: 165544471-165544471 on Build GRCh38
rs761360700 Chr.1: 165544466-165544466 on Build GRCh38
rs1050053795 Chr.1: 165544334-165544334 on Build GRCh38
rs529991653 Chr.1: 165544480-165544480 on Build GRCh38
rs771817384 Chr.1: 165544452-165544452 on Build GRCh38
rs774158608 Chr.1: 165544382-165544382 on Build GRCh38
rs760411659 Chr.1: 165544384-165544387 on Build GRCh38
rs745668125 Chr.1: 165544451-165544451 on Build GRCh38

View Full Product Details