Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 27344734-27344973 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL4R
Entrez Gene ID
3566
Gene Name
interleukin 4 receptor
Gene Aliases
CD124, IL-4RA, IL4RA
UniGene
Hs.513457
Gene Chromosome Location
Chr.16: 27313668-27364778 on Build GRCh38
Transcripts
NM_000418.3 NM_001257406.1 NM_001257407.1 NM_001257997.1 XM_011545825.1 XM_011545826.2 XM_011545827.2 XM_011545828.2 XM_011545829.1 XM_011545830.2 XM_011545831.1 XM_011545832.1 XM_011545833.2 XM_011545834.2 XM_017023211.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5619987 c.232A>T p.N78Y Chr.16: 27344891-27344891 on Build GRCh38
1210599 c.218C>T p.T73M Chr.16: 27344877-27344877 on Build GRCh38
6294584 c.253G>T p.V85L Chr.16: 27344912-27344912 on Build GRCh38
3108715 c.243C>T p.G81G Chr.16: 27344902-27344902 on Build GRCh38
6685414 c.277G>A p.V93M Chr.16: 27344936-27344936 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs554970103 Chr.16: 27344857-27344857 on Build GRCh38
rs761781246 Chr.16: 27344952-27344952 on Build GRCh38
rs893287811 Chr.16: 27344816-27344816 on Build GRCh38
rs375159221 Chr.16: 27344921-27344921 on Build GRCh38
rs1805010 Chr.16: 27344882-27344882 on Build GRCh38
rs145184963 Chr.16: 27344896-27344896 on Build GRCh38
rs752792692 Chr.16: 27344909-27344909 on Build GRCh38
rs750194290 Chr.16: 27344904-27344904 on Build GRCh38
rs113886028 Chr.16: 27344862-27344862 on Build GRCh38
rs141983833 Chr.16: 27344912-27344912 on Build GRCh38
rs144651842 Chr.16: 27344903-27344903 on Build GRCh38
rs781298536 Chr.16: 27344906-27344906 on Build GRCh38
rs759162058 Chr.16: 27344880-27344880 on Build GRCh38
rs527732961 Chr.16: 27344760-27344760 on Build GRCh38
rs775052929 Chr.16: 27344942-27344942 on Build GRCh38
rs775017923 Chr.16: 27344887-27344887 on Build GRCh38
rs1035376706 Chr.16: 27344879-27344879 on Build GRCh38
rs770243204 Chr.16: 27344826-27344826 on Build GRCh38
rs902076661 Chr.16: 27344784-27344784 on Build GRCh38
rs1022034958 Chr.16: 27344856-27344856 on Build GRCh38
rs569547141 Chr.16: 27344878-27344878 on Build GRCh38
rs996376416 Chr.16: 27344803-27344803 on Build GRCh38
rs940695158 Chr.16: 27344771-27344771 on Build GRCh38
rs141228265 Chr.16: 27344758-27344759 on Build GRCh38
rs367548039 Chr.16: 27344902-27344902 on Build GRCh38
rs748195646 Chr.16: 27344819-27344819 on Build GRCh38
rs190162514 Chr.16: 27344761-27344761 on Build GRCh38
rs999545268 Chr.16: 27344817-27344817 on Build GRCh38
rs530143808 Chr.16: 27344804-27344804 on Build GRCh38
rs764099093 Chr.16: 27344897-27344897 on Build GRCh38
rs763997764 Chr.16: 27344755-27344755 on Build GRCh38
rs755176173 Chr.16: 27344905-27344905 on Build GRCh38
rs895316248 Chr.16: 27344790-27344790 on Build GRCh38
rs778294946 Chr.16: 27344834-27344834 on Build GRCh38
rs565258570 Chr.16: 27344754-27344754 on Build GRCh38
rs746241652 Chr.16: 27344935-27344935 on Build GRCh38
rs902582750 Chr.16: 27344809-27344809 on Build GRCh38
rs778243651 Chr.16: 27344916-27344916 on Build GRCh38
rs112497527 Chr.16: 27344950-27344950 on Build GRCh38
rs368631624 Chr.16: 27344926-27344926 on Build GRCh38
rs1010665730 Chr.16: 27344833-27344833 on Build GRCh38
rs760218604 Chr.16: 27344946-27344946 on Build GRCh38
rs745518329 Chr.16: 27344859-27344859 on Build GRCh38
rs1011903425 Chr.16: 27344890-27344890 on Build GRCh38
rs771717750 Chr.16: 27344936-27344936 on Build GRCh38
rs372171065 Chr.16: 27344917-27344917 on Build GRCh38
rs907951146 Chr.16: 27344907-27344907 on Build GRCh38
rs949504759 Chr.16: 27344798-27344798 on Build GRCh38
rs567712475 Chr.16: 27344783-27344783 on Build GRCh38

View Full Product Details