Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 49543824-49544063 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNH3
Entrez Gene ID
23416
Gene Name
potassium voltage-gated channel subfamily H member 3
Gene Aliases
BEC1, ELK2, Kv12.2
UniGene
Hs.64064
Gene Chromosome Location
Chr.12: 49538541-49558312 on Build GRCh38
Transcripts
NM_001314030.1 NM_012284.2 XM_011538085.2 XM_011538086.2 XM_017019096.1 XM_017019097.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6345367 c.907G>T p.V303F Chr.12: 49543998-49543998 on Build GRCh38
431233 c.842G>A p.R281H Chr.12: 49543933-49543933 on Build GRCh38
6690906 c.904T>C p.Y302H Chr.12: 49543995-49543995 on Build GRCh38
7092661 c.852C>T p.F284F Chr.12: 49543943-49543943 on Build GRCh38
4147123 c.939A>G p.A313A Chr.12: 49544030-49544030 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781395888 Chr.12: 49543887-49543887 on Build GRCh38
rs779839329 Chr.12: 49543874-49543874 on Build GRCh38
rs746712922 Chr.12: 49543872-49543873 on Build GRCh38
rs200183735 Chr.12: 49543872-49543872 on Build GRCh38
rs768140928 Chr.12: 49543879-49543879 on Build GRCh38
rs754653406 Chr.12: 49543970-49543970 on Build GRCh38
rs879180240 Chr.12: 49543870-49543871 on Build GRCh38
rs1044785166 Chr.12: 49543903-49543903 on Build GRCh38
rs749110669 Chr.12: 49543875-49543875 on Build GRCh38
rs770013881 Chr.12: 49543901-49543901 on Build GRCh38
rs778808863 Chr.12: 49543988-49543988 on Build GRCh38
rs144378067 Chr.12: 49544033-49544033 on Build GRCh38
rs538981376 Chr.12: 49543916-49543916 on Build GRCh38
rs750007551 Chr.12: 49543954-49543954 on Build GRCh38
rs777509516 Chr.12: 49544027-49544027 on Build GRCh38
rs534065563 Chr.12: 49543855-49543855 on Build GRCh38
rs869167854 Chr.12: 49543878-49543878 on Build GRCh38
rs762426010 Chr.12: 49543937-49543937 on Build GRCh38
rs771872673 Chr.12: 49543879-49543879 on Build GRCh38
rs554063965 Chr.12: 49543859-49543859 on Build GRCh38
rs752429985 Chr.12: 49543997-49543997 on Build GRCh38
rs371023037 Chr.12: 49543910-49543910 on Build GRCh38
rs376939446 Chr.12: 49543877-49543877 on Build GRCh38
rs748165905 Chr.12: 49543878-49543878 on Build GRCh38
rs570669129 Chr.12: 49543876-49543876 on Build GRCh38
rs755864524 Chr.12: 49543873-49543873 on Build GRCh38
rs768962980 Chr.12: 49543920-49543920 on Build GRCh38
rs745918867 Chr.12: 49543893-49543893 on Build GRCh38
rs1009797822 Chr.12: 49544034-49544034 on Build GRCh38
rs932411413 Chr.12: 49543974-49543974 on Build GRCh38
rs750741267 Chr.12: 49543967-49543967 on Build GRCh38
rs760207096 Chr.12: 49543955-49543955 on Build GRCh38
rs1000470759 Chr.12: 49544015-49544015 on Build GRCh38
rs11337899 Chr.12: 49543872-49543872 on Build GRCh38
rs558683094 Chr.12: 49543933-49543933 on Build GRCh38
rs762725272 Chr.12: 49543871-49543872 on Build GRCh38
rs913822106 Chr.12: 49543904-49543904 on Build GRCh38
rs758415878 Chr.12: 49544000-49544000 on Build GRCh38
rs767907185 Chr.12: 49543943-49543943 on Build GRCh38
rs1150057 Chr.12: 49544030-49544030 on Build GRCh38
rs775620693 Chr.12: 49543905-49543905 on Build GRCh38
rs753490202 Chr.12: 49543961-49543961 on Build GRCh38
rs776267384 Chr.12: 49543879-49543880 on Build GRCh38

View Full Product Details