Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 6624805-6625076 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LY86
Entrez Gene ID
9450
Gene Name
lymphocyte antigen 86
Gene Aliases
MD-1, MD1, MMD-1, dJ80N2.1
UniGene
Hs.653138
Gene Chromosome Location
Chr.6: 6588639-6654983 on Build GRCh38
Transcripts
NM_004271.3 XM_017011505.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6843483 c.173G>C p.C58S Chr.6: 6624962-6624962 on Build GRCh38
3160701 c.139C>T p.P47S Chr.6: 6624928-6624928 on Build GRCh38
6207994 c.196A>T p.I66F Chr.6: 6624985-6624985 on Build GRCh38
4432782 c.181C>A p.Q61K Chr.6: 6624970-6624970 on Build GRCh38
3725739 c.137-5C>T p.? Chr.6: 6624921-6624921 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754994838 Chr.6: 6625010-6625013 on Build GRCh38
rs201924366 Chr.6: 6624963-6624963 on Build GRCh38
rs749837341 Chr.6: 6624887-6624887 on Build GRCh38
rs747701104 Chr.6: 6624966-6624966 on Build GRCh38
rs768219674 Chr.6: 6624880-6624880 on Build GRCh38
rs370252261 Chr.6: 6624922-6624922 on Build GRCh38
rs779650098 Chr.6: 6624955-6624955 on Build GRCh38
rs113353455 Chr.6: 6624886-6624886 on Build GRCh38
rs553977919 Chr.6: 6625028-6625028 on Build GRCh38
rs200582242 Chr.6: 6625007-6625007 on Build GRCh38
rs753964742 Chr.6: 6624923-6624923 on Build GRCh38
rs1010249798 Chr.6: 6624884-6624884 on Build GRCh38
rs750318701 Chr.6: 6624939-6624939 on Build GRCh38
rs569300613 Chr.6: 6624992-6624992 on Build GRCh38
rs761847184 Chr.6: 6624935-6624935 on Build GRCh38
rs761935131 Chr.6: 6625041-6625041 on Build GRCh38
rs772516616 Chr.6: 6625010-6625010 on Build GRCh38
rs954091736 Chr.6: 6624989-6624989 on Build GRCh38
rs371700147 Chr.6: 6624875-6624875 on Build GRCh38
rs576973543 Chr.6: 6625054-6625054 on Build GRCh38
rs760012549 Chr.6: 6624904-6624904 on Build GRCh38
rs148799324 Chr.6: 6624952-6624952 on Build GRCh38
rs955987499 Chr.6: 6624850-6624850 on Build GRCh38
rs142377958 Chr.6: 6625004-6625004 on Build GRCh38
rs765247947 Chr.6: 6625049-6625049 on Build GRCh38
rs34247656 Chr.6: 6624955-6624956 on Build GRCh38
rs751259303 Chr.6: 6624956-6624956 on Build GRCh38
rs891194737 Chr.6: 6624847-6624847 on Build GRCh38
rs775784697 Chr.6: 6624921-6624921 on Build GRCh38
rs780971858 Chr.6: 6624965-6624965 on Build GRCh38
rs779172172 Chr.6: 6625019-6625019 on Build GRCh38
rs768904537 Chr.6: 6624946-6624946 on Build GRCh38
rs772267003 Chr.6: 6624909-6624909 on Build GRCh38
rs771616229 Chr.6: 6624895-6624895 on Build GRCh38
rs775666814 Chr.6: 6625019-6625019 on Build GRCh38
rs762763398 Chr.6: 6625055-6625055 on Build GRCh38
rs761075518 Chr.6: 6625034-6625034 on Build GRCh38
rs372994760 Chr.6: 6624896-6624896 on Build GRCh38
rs762733808 Chr.6: 6625033-6625033 on Build GRCh38
rs765221544 Chr.6: 6624936-6624936 on Build GRCh38
rs866108570 Chr.6: 6624950-6624950 on Build GRCh38
rs1029847661 Chr.6: 6624845-6624845 on Build GRCh38
rs748354471 Chr.6: 6625018-6625019 on Build GRCh38
rs376096581 Chr.6: 6625035-6625035 on Build GRCh38
rs776826225 Chr.6: 6625040-6625040 on Build GRCh38

View Full Product Details