Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 108070656-108070909 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LEF1
Entrez Gene ID
51176
Gene Name
lymphoid enhancer binding factor 1
Gene Aliases
LEF-1, TCF10, TCF1ALPHA, TCF7L3
UniGene
Hs.743478
Gene Chromosome Location
Chr.4: 108047545-108168956 on Build GRCh38
Transcripts
NM_001130713.2 NM_001130714.2 NM_001166119.1 NM_016269.4 XM_005263046.3 XM_005263047.1 XM_005263048.1 XM_006714233.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6165859 c.1094G>T p.G365V Chr.4: 108070685-108070685 on Build GRCh38
6165861 c.806G>T p.G269V Chr.4: 108070685-108070685 on Build GRCh38
7297982 c.824A>G p.N275S Chr.4: 108070667-108070667 on Build GRCh38
3380778 c.769C>T p.R257W Chr.4: 108070722-108070722 on Build GRCh38
6165856 c.1025G>T p.R342L Chr.4: 108070754-108070754 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148125388 Chr.4: 108070726-108070726 on Build GRCh38
rs200011152 Chr.4: 108070754-108070754 on Build GRCh38
rs147753559 Chr.4: 108070756-108070756 on Build GRCh38
rs753999750 Chr.4: 108070705-108070705 on Build GRCh38
rs765332289 Chr.4: 108070820-108070820 on Build GRCh38
rs765557471 Chr.4: 108070723-108070723 on Build GRCh38
rs760616868 Chr.4: 108070808-108070808 on Build GRCh38
rs745539236 Chr.4: 108070861-108070861 on Build GRCh38
rs764159891 Chr.4: 108070696-108070696 on Build GRCh38
rs1032490478 Chr.4: 108070715-108070715 on Build GRCh38
rs768707372 Chr.4: 108070809-108070809 on Build GRCh38
rs557145182 Chr.4: 108070832-108070832 on Build GRCh38
rs766916120 Chr.4: 108070711-108070711 on Build GRCh38
rs780385841 Chr.4: 108070733-108070733 on Build GRCh38
rs773519468 Chr.4: 108070821-108070821 on Build GRCh38
rs141886471 Chr.4: 108070806-108070806 on Build GRCh38
rs771928491 Chr.4: 108070805-108070805 on Build GRCh38
rs778745857 Chr.4: 108070772-108070772 on Build GRCh38
rs770745649 Chr.4: 108070766-108070766 on Build GRCh38
rs574079007 Chr.4: 108070855-108070855 on Build GRCh38
rs745600382 Chr.4: 108070799-108070799 on Build GRCh38
rs748942249 Chr.4: 108070759-108070759 on Build GRCh38
rs752111922 Chr.4: 108070747-108070747 on Build GRCh38
rs2276335 Chr.4: 108070823-108070823 on Build GRCh38
rs779619674 Chr.4: 108070787-108070787 on Build GRCh38
rs369312312 Chr.4: 108070816-108070816 on Build GRCh38
rs760702658 Chr.4: 108070685-108070685 on Build GRCh38
rs758817161 Chr.4: 108070732-108070732 on Build GRCh38

View Full Product Details