Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 73197207-73197434 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COG1
Entrez Gene ID
9382
Gene Name
component of oligomeric golgi complex 1
Gene Aliases
CDG2G, LDLB
UniGene
Hs.103555
Gene Chromosome Location
Chr.17: 73193034-73208507 on Build GRCh38
Transcripts
NM_018714.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7415780 c.880A>T p.T294S Chr.17: 73197363-73197363 on Build GRCh38
5501140 c.847G>A p.D283N Chr.17: 73197330-73197330 on Build GRCh38
7361908 c.753C>G p.I251M Chr.17: 73197236-73197236 on Build GRCh38
5916573 c.913+8C>T p.? Chr.17: 73197404-73197404 on Build GRCh38
2976469 c.814C>A p.L272I Chr.17: 73197297-73197297 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772932593 Chr.17: 73197314-73197314 on Build GRCh38
rs767482114 Chr.17: 73197345-73197345 on Build GRCh38
rs769317853 Chr.17: 73197311-73197311 on Build GRCh38
rs759447552 Chr.17: 73197344-73197344 on Build GRCh38
rs139304974 Chr.17: 73197258-73197258 on Build GRCh38
rs750736176 Chr.17: 73197259-73197259 on Build GRCh38
rs750659961 Chr.17: 73197390-73197390 on Build GRCh38
rs747727663 Chr.17: 73197305-73197305 on Build GRCh38
rs766720884 Chr.17: 73197314-73197316 on Build GRCh38
rs776685074 Chr.17: 73197383-73197383 on Build GRCh38
rs747543891 Chr.17: 73197290-73197290 on Build GRCh38
rs147973539 Chr.17: 73197395-73197395 on Build GRCh38
rs140691967 Chr.17: 73197321-73197321 on Build GRCh38
rs367884440 Chr.17: 73197274-73197274 on Build GRCh38
rs572583949 Chr.17: 73197361-73197361 on Build GRCh38
rs11919 Chr.17: 73197260-73197260 on Build GRCh38
rs751797257 Chr.17: 73197405-73197407 on Build GRCh38
rs117208167 Chr.17: 73197386-73197386 on Build GRCh38
rs755596131 Chr.17: 73197293-73197293 on Build GRCh38
rs374160707 Chr.17: 73197396-73197396 on Build GRCh38
rs770789819 Chr.17: 73197322-73197322 on Build GRCh38
rs774105621 Chr.17: 73197337-73197337 on Build GRCh38
rs780388109 Chr.17: 73197276-73197276 on Build GRCh38
rs755540810 Chr.17: 73197404-73197404 on Build GRCh38
rs376078151 Chr.17: 73197233-73197233 on Build GRCh38
rs780821062 Chr.17: 73197297-73197297 on Build GRCh38
rs757438614 Chr.17: 73197251-73197251 on Build GRCh38
rs752014690 Chr.17: 73197397-73197397 on Build GRCh38

View Full Product Details