Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 21249615-21249878 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKS4B
Entrez Gene ID
257629
Gene Name
ankyrin repeat and sterile alpha motif domain containing 4B
Gene Aliases
HARP
UniGene
Hs.115959
Gene Chromosome Location
Chr.16: 21233695-21252429 on Build GRCh38
Transcripts
NM_145865.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4059155 c.241G>A p.V81I Chr.16: 21249807-21249807 on Build GRCh38
4917908 c.244T>C p.S82P Chr.16: 21249810-21249810 on Build GRCh38
4885020 c.202C>A p.P68T Chr.16: 21249768-21249768 on Build GRCh38
5850428 c.253G>A p.V85I Chr.16: 21249819-21249819 on Build GRCh38
1182985 c.224A>G p.N75S Chr.16: 21249790-21249790 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746962210 Chr.16: 21249712-21249712 on Build GRCh38
rs372062197 Chr.16: 21249806-21249806 on Build GRCh38
rs754756882 Chr.16: 21249807-21249807 on Build GRCh38
rs750022603 Chr.16: 21249684-21249684 on Build GRCh38
rs749431930 Chr.16: 21249718-21249718 on Build GRCh38
rs1033581915 Chr.16: 21249710-21249710 on Build GRCh38
rs768765908 Chr.16: 21249745-21249745 on Build GRCh38
rs367892200 Chr.16: 21249782-21249782 on Build GRCh38
rs759633281 Chr.16: 21249746-21249746 on Build GRCh38
rs922185948 Chr.16: 21249825-21249825 on Build GRCh38
rs201709712 Chr.16: 21249771-21249771 on Build GRCh38
rs748049990 Chr.16: 21249815-21249815 on Build GRCh38
rs758252199 Chr.16: 21249694-21249694 on Build GRCh38
rs778684089 Chr.16: 21249681-21249681 on Build GRCh38
rs778804642 Chr.16: 21249835-21249835 on Build GRCh38
rs757930770 Chr.16: 21249796-21249796 on Build GRCh38
rs201323707 Chr.16: 21249713-21249714 on Build GRCh38
rs770897592 Chr.16: 21249720-21249720 on Build GRCh38
rs746792050 Chr.16: 21249706-21249706 on Build GRCh38
rs1010509288 Chr.16: 21249664-21249664 on Build GRCh38
rs891612868 Chr.16: 21249663-21249664 on Build GRCh38
rs779830541 Chr.16: 21249799-21249799 on Build GRCh38
rs1020973702 Chr.16: 21249668-21249668 on Build GRCh38
rs537428019 Chr.16: 21249695-21249695 on Build GRCh38
rs761434899 Chr.16: 21249783-21249783 on Build GRCh38
rs764621269 Chr.16: 21249784-21249784 on Build GRCh38
rs764763053 Chr.16: 21249781-21249781 on Build GRCh38
rs987400213 Chr.16: 21249678-21249678 on Build GRCh38
rs373793273 Chr.16: 21249703-21249703 on Build GRCh38
rs750113646 Chr.16: 21249790-21249790 on Build GRCh38
rs954807829 Chr.16: 21249672-21249672 on Build GRCh38
rs761266418 Chr.16: 21249774-21249774 on Build GRCh38
rs775569181 Chr.16: 21249687-21249687 on Build GRCh38
rs772504457 Chr.16: 21249768-21249768 on Build GRCh38
rs781159145 Chr.16: 21249810-21249810 on Build GRCh38
rs746111689 Chr.16: 21249844-21249844 on Build GRCh38
rs781104530 Chr.16: 21249717-21249717 on Build GRCh38
rs768629428 Chr.16: 21249705-21249706 on Build GRCh38
rs772115947 Chr.16: 21249856-21249856 on Build GRCh38
rs75187531 Chr.16: 21249824-21249824 on Build GRCh38
rs768515598 Chr.16: 21249714-21249714 on Build GRCh38
rs774678647 Chr.16: 21249744-21249744 on Build GRCh38
rs537864105 Chr.16: 21249850-21249850 on Build GRCh38

View Full Product Details