Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 38462869-38463134 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP23
Entrez Gene ID
57636
Gene Name
Rho GTPase activating protein 23
Gene Aliases
-
UniGene
Hs.374446
Gene Chromosome Location
Chr.17: 38419286-38512392 on Build GRCh38
Transcripts
NM_001199417.1 XM_006721991.1 XM_006721992.3 XM_006721993.3 XM_006721994.3 XM_011525073.1 XM_011525074.2 XM_011525076.2 XM_011525077.2 XM_017024902.1

SNP Detail

SNP ID Chromosome Location
rs866254733 Chr.17: 38462911-38462911 on Build GRCh38
rs145184951 Chr.17: 38463109-38463109 on Build GRCh38
rs959209681 Chr.17: 38463076-38463076 on Build GRCh38
rs1003300980 Chr.17: 38463057-38463057 on Build GRCh38
rs201978800 Chr.17: 38462935-38462935 on Build GRCh38
rs888055836 Chr.17: 38462945-38462945 on Build GRCh38
rs61999279 Chr.17: 38462898-38462898 on Build GRCh38
rs759443824 Chr.17: 38462957-38462957 on Build GRCh38
rs991034106 Chr.17: 38463093-38463093 on Build GRCh38
rs761587380 Chr.17: 38463074-38463074 on Build GRCh38
rs752943213 Chr.17: 38463069-38463069 on Build GRCh38
rs1054520339 Chr.17: 38462964-38462964 on Build GRCh38
rs763963292 Chr.17: 38462984-38462984 on Build GRCh38
rs554567775 Chr.17: 38463102-38463102 on Build GRCh38
rs765125715 Chr.17: 38462966-38462966 on Build GRCh38
rs149987232 Chr.17: 38462960-38462960 on Build GRCh38
rs767978482 Chr.17: 38463051-38463051 on Build GRCh38
rs949980160 Chr.17: 38462916-38462916 on Build GRCh38
rs900687243 Chr.17: 38462920-38462920 on Build GRCh38
rs762712770 Chr.17: 38462978-38462978 on Build GRCh38
rs753160587 Chr.17: 38463048-38463048 on Build GRCh38
rs538668316 Chr.17: 38462910-38462910 on Build GRCh38
rs146555807 Chr.17: 38462985-38462985 on Build GRCh38
rs932113153 Chr.17: 38462928-38462928 on Build GRCh38
rs1015713682 Chr.17: 38462973-38462973 on Build GRCh38
rs775347460 Chr.17: 38462968-38462968 on Build GRCh38
rs866797645 Chr.17: 38463092-38463092 on Build GRCh38
rs1034726903 Chr.17: 38463070-38463070 on Build GRCh38

View Full Product Details