Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 76491434-76491689 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ESRRB
Entrez Gene ID
2103
Gene Name
estrogen related receptor beta
Gene Aliases
DFNB35, ERR2, ERRb, ESRL2, NR3B2
UniGene
Hs.435845
Gene Chromosome Location
Chr.14: 76371347-76501839 on Build GRCh38
Transcripts
NM_004452.3 XM_011536547.2 XM_011536550.2 XM_011536553.2 XM_011536554.2 XM_017021085.1 XM_017021086.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3956227 c.839G>T p.W280L Chr.14: 76491498-76491498 on Build GRCh38
6434528 c.875C>T p.S292L Chr.14: 76491534-76491534 on Build GRCh38
4567661 c.807_808GG>AA p.G270R Chr.14: 76491466-76491467 on Build GRCh38
6403141 c.938G>A p.R313H Chr.14: 76491597-76491597 on Build GRCh38
4559606 c.811G>A p.D271N Chr.14: 76491470-76491470 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs576301680 Chr.14: 76491575-76491575 on Build GRCh38
rs754479043 Chr.14: 76491458-76491458 on Build GRCh38
rs991653082 Chr.14: 76491534-76491534 on Build GRCh38
rs750798786 Chr.14: 76491531-76491531 on Build GRCh38
rs528588232 Chr.14: 76491645-76491645 on Build GRCh38
rs777504452 Chr.14: 76491654-76491654 on Build GRCh38
rs767567875 Chr.14: 76491617-76491617 on Build GRCh38
rs766812921 Chr.14: 76491627-76491627 on Build GRCh38
rs899007798 Chr.14: 76491634-76491634 on Build GRCh38
rs193189009 Chr.14: 76491586-76491586 on Build GRCh38
rs749055650 Chr.14: 76491669-76491669 on Build GRCh38
rs887112300 Chr.14: 76491457-76491457 on Build GRCh38
rs369987440 Chr.14: 76491559-76491559 on Build GRCh38
rs764181534 Chr.14: 76491603-76491603 on Build GRCh38
rs745788988 Chr.14: 76491576-76491576 on Build GRCh38
rs764276965 Chr.14: 76491515-76491515 on Build GRCh38
rs876657463 Chr.14: 76491602-76491602 on Build GRCh38
rs765391919 Chr.14: 76491481-76491481 on Build GRCh38
rs143118664 Chr.14: 76491623-76491623 on Build GRCh38
rs754105207 Chr.14: 76491644-76491644 on Build GRCh38
rs1007989619 Chr.14: 76491453-76491453 on Build GRCh38
rs757790651 Chr.14: 76491523-76491523 on Build GRCh38
rs767944860 Chr.14: 76491524-76491524 on Build GRCh38
rs760746815 Chr.14: 76491456-76491456 on Build GRCh38
rs139812473 Chr.14: 76491547-76491547 on Build GRCh38
rs775118593 Chr.14: 76491596-76491596 on Build GRCh38
rs775840099 Chr.14: 76491483-76491483 on Build GRCh38
rs145982062 Chr.14: 76491545-76491545 on Build GRCh38
rs2361293 Chr.14: 76491544-76491544 on Build GRCh38
rs756555664 Chr.14: 76491535-76491535 on Build GRCh38
rs201417586 Chr.14: 76491472-76491472 on Build GRCh38
rs780275423 Chr.14: 76491543-76491543 on Build GRCh38
rs780688931 Chr.14: 76491478-76491478 on Build GRCh38
rs201344770 Chr.14: 76491548-76491548 on Build GRCh38
rs761822998 Chr.14: 76491616-76491616 on Build GRCh38
rs886038549 Chr.14: 76491628-76491628 on Build GRCh38
rs267604057 Chr.14: 76491502-76491502 on Build GRCh38
rs146185697 Chr.14: 76491624-76491624 on Build GRCh38
rs763083614 Chr.14: 76491485-76491485 on Build GRCh38
rs769709962 Chr.14: 76491572-76491572 on Build GRCh38
rs1032046141 Chr.14: 76491490-76491490 on Build GRCh38
rs771001016 Chr.14: 76491562-76491562 on Build GRCh38
rs774418850 Chr.14: 76491604-76491604 on Build GRCh38
rs776652286 Chr.14: 76491563-76491563 on Build GRCh38
rs148356050 Chr.14: 76491520-76491520 on Build GRCh38
rs981693942 Chr.14: 76491550-76491550 on Build GRCh38
rs763256376 Chr.14: 76491597-76491597 on Build GRCh38

View Full Product Details