Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 69236336-69236609 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXD2
Entrez Gene ID
55218
Gene Name
exonuclease 3'-5' domain containing 2
Gene Aliases
C14orf114, EXDL2
UniGene
Hs.533878
Gene Chromosome Location
Chr.14: 69191477-69244020 on Build GRCh38
Transcripts
NM_001193360.1 NM_001193361.1 NM_001193362.1 NM_001193363.1 NM_018199.3 NR_034164.1 NR_034165.1 XM_005267817.3 XM_011536908.2 XM_017021421.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1370776 c.909C>T p.S303S Chr.14: 69236534-69236534 on Build GRCh38
5618742 c.816A>G p.L272L Chr.14: 69236441-69236441 on Build GRCh38
4798658 c.1212G>A p.R404R Chr.14: 69236462-69236462 on Build GRCh38
6657020 c.840C>T p.P280P Chr.14: 69236465-69236465 on Build GRCh38
1743283 c.1192C>T p.R398W Chr.14: 69236442-69236442 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370664411 Chr.14: 69236568-69236568 on Build GRCh38
rs770410901 Chr.14: 69236360-69236360 on Build GRCh38
rs937573986 Chr.14: 69236365-69236365 on Build GRCh38
rs566133114 Chr.14: 69236443-69236443 on Build GRCh38
rs540160942 Chr.14: 69236465-69236465 on Build GRCh38
rs1029551609 Chr.14: 69236555-69236555 on Build GRCh38
rs762688092 Chr.14: 69236393-69236393 on Build GRCh38
rs764429667 Chr.14: 69236580-69236580 on Build GRCh38
rs761102147 Chr.14: 69236524-69236525 on Build GRCh38
rs755346202 Chr.14: 69236475-69236475 on Build GRCh38
rs369000595 Chr.14: 69236395-69236395 on Build GRCh38
rs776001428 Chr.14: 69236399-69236399 on Build GRCh38
rs3211166 Chr.14: 69236441-69236441 on Build GRCh38
rs570334436 Chr.14: 69236544-69236544 on Build GRCh38
rs542694805 Chr.14: 69236507-69236507 on Build GRCh38
rs373719960 Chr.14: 69236542-69236542 on Build GRCh38
rs200631654 Chr.14: 69236482-69236482 on Build GRCh38
rs141991626 Chr.14: 69236541-69236541 on Build GRCh38
rs199623062 Chr.14: 69236561-69236561 on Build GRCh38
rs760671586 Chr.14: 69236557-69236557 on Build GRCh38
rs867718202 Chr.14: 69236476-69236476 on Build GRCh38
rs771623552 Chr.14: 69236581-69236581 on Build GRCh38
rs759817723 Chr.14: 69236566-69236566 on Build GRCh38
rs138356811 Chr.14: 69236547-69236547 on Build GRCh38
rs775310562 Chr.14: 69236385-69236385 on Build GRCh38
rs146968535 Chr.14: 69236466-69236466 on Build GRCh38
rs147040929 Chr.14: 69236363-69236363 on Build GRCh38
rs983019098 Chr.14: 69236579-69236579 on Build GRCh38
rs774444788 Chr.14: 69236409-69236409 on Build GRCh38
rs773301644 Chr.14: 69236548-69236548 on Build GRCh38
rs745504721 Chr.14: 69236367-69236367 on Build GRCh38
rs769385275 Chr.14: 69236371-69236371 on Build GRCh38
rs552080663 Chr.14: 69236526-69236526 on Build GRCh38
rs780735861 Chr.14: 69236505-69236505 on Build GRCh38
rs933450722 Chr.14: 69236525-69236525 on Build GRCh38
rs371436325 Chr.14: 69236583-69236583 on Build GRCh38
rs779305241 Chr.14: 69236481-69236481 on Build GRCh38
rs745396157 Chr.14: 69236524-69236524 on Build GRCh38
rs771502862 Chr.14: 69236393-69236397 on Build GRCh38
rs922181403 Chr.14: 69236515-69236515 on Build GRCh38
rs147562168 Chr.14: 69236491-69236491 on Build GRCh38
rs753991134 Chr.14: 69236563-69236563 on Build GRCh38
rs758946926 Chr.14: 69236500-69236500 on Build GRCh38
rs761901916 Chr.14: 69236429-69236429 on Build GRCh38

View Full Product Details