Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 48434594-48434865 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC51
Entrez Gene ID
79714
Gene Name
coiled-coil domain containing 51
Gene Aliases
-
UniGene
Hs.187657
Gene Chromosome Location
Chr.3: 48432170-48446652 on Build GRCh38
Transcripts
NM_001256964.1 NM_001256965.2 NM_001256966.2 NM_001256967.2 NM_001256968.2 NM_001256969.2 NM_024661.4 XM_011534113.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6622248 c.283G>A p.V95I Chr.3: 48434846-48434846 on Build GRCh38
1328182 c.278A>G p.N93S Chr.3: 48434851-48434851 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs960274619 Chr.3: 48434636-48434636 on Build GRCh38
rs749100598 Chr.3: 48434703-48434703 on Build GRCh38
rs546564076 Chr.3: 48434786-48434786 on Build GRCh38
rs748599525 Chr.3: 48434787-48434787 on Build GRCh38
rs565861189 Chr.3: 48434797-48434797 on Build GRCh38
rs879102322 Chr.3: 48434740-48434740 on Build GRCh38
rs749406049 Chr.3: 48434798-48434798 on Build GRCh38
rs986634599 Chr.3: 48434777-48434777 on Build GRCh38
rs191317794 Chr.3: 48434807-48434807 on Build GRCh38
rs746361275 Chr.3: 48434702-48434702 on Build GRCh38
rs139486669 Chr.3: 48434657-48434657 on Build GRCh38
rs755335976 Chr.3: 48434784-48434784 on Build GRCh38
rs1024562595 Chr.3: 48434662-48434662 on Build GRCh38
rs867746959 Chr.3: 48434726-48434726 on Build GRCh38
rs759904380 Chr.3: 48434804-48434804 on Build GRCh38
rs560356888 Chr.3: 48434743-48434743 on Build GRCh38
rs372223794 Chr.3: 48434793-48434793 on Build GRCh38
rs934748874 Chr.3: 48434729-48434729 on Build GRCh38
rs1049159928 Chr.3: 48434637-48434637 on Build GRCh38
rs566054941 Chr.3: 48434705-48434705 on Build GRCh38
rs773122337 Chr.3: 48434795-48434795 on Build GRCh38
rs907490507 Chr.3: 48434699-48434699 on Build GRCh38
rs911928544 Chr.3: 48434762-48434762 on Build GRCh38
rs957076874 Chr.3: 48434736-48434736 on Build GRCh38
rs374407462 Chr.3: 48434799-48434799 on Build GRCh38
rs186408202 Chr.3: 48434644-48434644 on Build GRCh38
rs368487934 Chr.3: 48434767-48434767 on Build GRCh38
rs756134469 Chr.3: 48434650-48434650 on Build GRCh38
rs200464374 Chr.3: 48434842-48434842 on Build GRCh38
rs916047678 Chr.3: 48434645-48434645 on Build GRCh38
rs775436647 Chr.3: 48434843-48434843 on Build GRCh38
rs879028982 Chr.3: 48434741-48434741 on Build GRCh38
rs774679864 Chr.3: 48434800-48434800 on Build GRCh38
rs527586566 Chr.3: 48434751-48434751 on Build GRCh38

View Full Product Details