Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 101476111-101476344 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM246
Entrez Gene ID
84302
Gene Name
transmembrane protein 246
Gene Aliases
C9orf125
UniGene
Hs.388742
Gene Chromosome Location
Chr.9: 101473158-101487221 on Build GRCh38
Transcripts
NM_001303107.1 NM_001303108.1 NM_032342.2 XM_017015206.1 NR_121573.1 NR_121574.1 NR_121575.1 NR_121576.1

Gene Symbol
TMEM246-AS1
Entrez Gene ID
101928470
Gene Name
TMEM246 antisense RNA 1
Gene Aliases
-
UniGene
Hs.656088
Gene Chromosome Location
Chr.9: 101468439-101481506 on Build GRCh38
Transcripts
NM_001303107.1 NM_001303108.1 NM_032342.2 XM_017015206.1 NR_121573.1 NR_121574.1 NR_121575.1 NR_121576.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3652380 c.789G>A p.L263L Chr.9: 101476304-101476304 on Build GRCh38
3925796 c.939C>T p.F313F Chr.9: 101476154-101476154 on Build GRCh38
752266 c.816G>T p.L272L Chr.9: 101476277-101476277 on Build GRCh38
3925797 c.783G>A p.R261R Chr.9: 101476310-101476310 on Build GRCh38
4643281 c.757C>T p.H253Y Chr.9: 101476336-101476336 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759069394 Chr.9: 101476274-101476276 on Build GRCh38
rs757079504 Chr.9: 101476218-101476218 on Build GRCh38
rs774714298 Chr.9: 101476251-101476251 on Build GRCh38
rs774847252 Chr.9: 101476282-101476282 on Build GRCh38
rs763250345 Chr.9: 101476143-101476143 on Build GRCh38
rs143415385 Chr.9: 101476185-101476185 on Build GRCh38
rs759924926 Chr.9: 101476295-101476295 on Build GRCh38
rs779898081 Chr.9: 101476243-101476243 on Build GRCh38
rs149643147 Chr.9: 101476141-101476141 on Build GRCh38
rs759438430 Chr.9: 101476271-101476271 on Build GRCh38
rs760121031 Chr.9: 101476161-101476161 on Build GRCh38
rs771493396 Chr.9: 101476280-101476280 on Build GRCh38
rs750417522 Chr.9: 101476236-101476236 on Build GRCh38
rs748209653 Chr.9: 101476247-101476247 on Build GRCh38
rs766828805 Chr.9: 101476144-101476144 on Build GRCh38
rs768374892 Chr.9: 101476248-101476248 on Build GRCh38
rs752166272 Chr.9: 101476171-101476171 on Build GRCh38
rs878933419 Chr.9: 101476297-101476297 on Build GRCh38
rs769913773 Chr.9: 101476137-101476137 on Build GRCh38
rs760189987 Chr.9: 101476172-101476172 on Build GRCh38
rs775942742 Chr.9: 101476268-101476268 on Build GRCh38
rs753501953 Chr.9: 101476213-101476213 on Build GRCh38
rs557639064 Chr.9: 101476142-101476142 on Build GRCh38
rs748200484 Chr.9: 101476136-101476136 on Build GRCh38
rs767196637 Chr.9: 101476163-101476163 on Build GRCh38
rs928790005 Chr.9: 101476252-101476252 on Build GRCh38
rs891808557 Chr.9: 101476314-101476314 on Build GRCh38
rs960904080 Chr.9: 101476139-101476139 on Build GRCh38
rs778673337 Chr.9: 101476219-101476219 on Build GRCh38
rs775041580 Chr.9: 101476156-101476156 on Build GRCh38
rs376517979 Chr.9: 101476237-101476237 on Build GRCh38
rs773446003 Chr.9: 101476140-101476140 on Build GRCh38
rs138246997 Chr.9: 101476132-101476132 on Build GRCh38

View Full Product Details