Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 244477799-244478072 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf101
Entrez Gene ID
257044
Gene Name
chromosome 1 open reading frame 101
Gene Aliases
-
UniGene
Hs.459534
Gene Chromosome Location
Chr.1: 244452014-244640362 on Build GRCh38
Transcripts
NM_001130957.1 NM_001242340.1 NM_173807.4 XM_011544138.2 XM_011544139.2 XM_011544142.2 XM_011544144.2 XM_011544145.2 XM_011544149.2 XM_011544151.2 XM_017000943.1 XM_017000944.1 XM_017000945.1 XM_017000946.1 XM_017000947.1 XM_017000948.1 XM_017000949.1 XM_017000950.1 XM_017000951.1 XM_017000952.1 XM_017000953.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4389191 c.232G>A p.A78T Chr.1: 244477949-244477949 on Build GRCh38
5481036 c.209G>A p.R70H Chr.1: 244477926-244477926 on Build GRCh38
5481037 c.209G>A p.R70H Chr.1: 244477926-244477926 on Build GRCh38
5502412 c.189-2A>T p.? Chr.1: 244477904-244477904 on Build GRCh38
3377080 c.222T>C p.P74P Chr.1: 244477939-244477939 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs988828279 Chr.1: 244478009-244478009 on Build GRCh38
rs200697066 Chr.1: 244477949-244477949 on Build GRCh38
rs993848588 Chr.1: 244477840-244477840 on Build GRCh38
rs763330537 Chr.1: 244478013-244478013 on Build GRCh38
rs117468858 Chr.1: 244477873-244477873 on Build GRCh38
rs368627637 Chr.1: 244477858-244477858 on Build GRCh38
rs374717406 Chr.1: 244477848-244477848 on Build GRCh38
rs745658494 Chr.1: 244477948-244477948 on Build GRCh38
rs776375193 Chr.1: 244477870-244477870 on Build GRCh38
rs376144649 Chr.1: 244477995-244477995 on Build GRCh38
rs112627425 Chr.1: 244478016-244478016 on Build GRCh38
rs769443325 Chr.1: 244477983-244477983 on Build GRCh38
rs766836074 Chr.1: 244478023-244478023 on Build GRCh38
rs144274959 Chr.1: 244477952-244477952 on Build GRCh38
rs775070062 Chr.1: 244477860-244477860 on Build GRCh38
rs761524748 Chr.1: 244477982-244477982 on Build GRCh38
rs778844324 Chr.1: 244477932-244477932 on Build GRCh38
rs760245310 Chr.1: 244477862-244477862 on Build GRCh38
rs559442266 Chr.1: 244477925-244477925 on Build GRCh38
rs746462117 Chr.1: 244477965-244477965 on Build GRCh38
rs200178961 Chr.1: 244477906-244477906 on Build GRCh38
rs189806894 Chr.1: 244477869-244477869 on Build GRCh38
rs763383657 Chr.1: 244477904-244477904 on Build GRCh38
rs768178913 Chr.1: 244477970-244477970 on Build GRCh38
rs776053074 Chr.1: 244477976-244477976 on Build GRCh38
rs914552373 Chr.1: 244478028-244478028 on Build GRCh38
rs202153521 Chr.1: 244477900-244477900 on Build GRCh38
rs11586356 Chr.1: 244477914-244477914 on Build GRCh38
rs781171580 Chr.1: 244477924-244477924 on Build GRCh38
rs924711444 Chr.1: 244477841-244477841 on Build GRCh38
rs947699471 Chr.1: 244478040-244478040 on Build GRCh38
rs765573729 Chr.1: 244477894-244477894 on Build GRCh38
rs115480880 Chr.1: 244477926-244477926 on Build GRCh38
rs956088335 Chr.1: 244477966-244477966 on Build GRCh38

View Full Product Details