Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 122980693-122980942 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL6IP4
Entrez Gene ID
51329
Gene Name
ADP ribosylation factor like GTPase 6 interacting protein 4
Gene Aliases
SFRS20, SR-25, SRp25, SRrp37
UniGene
Hs.103561
Gene Chromosome Location
Chr.12: 122980060-122982913 on Build GRCh38
Transcripts
NM_001002251.2 NM_001002252.2 NM_001278378.1 NM_001278379.1 NM_001278380.1 NM_016638.3 NM_018694.3 NR_103512.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4655197 c.331G>A p.G111R Chr.12: 122980718-122980718 on Build GRCh38
6380129 c.332G>T p.G111V Chr.12: 122980719-122980719 on Build GRCh38
5554491 c.351C>A p.G117G Chr.12: 122980738-122980738 on Build GRCh38
5554490 c.351C>A p.G117G Chr.12: 122980738-122980738 on Build GRCh38
7190306 c.358+1G>T p.? Chr.12: 122980746-122980746 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs967584474 Chr.12: 122980842-122980842 on Build GRCh38
rs953318008 Chr.12: 122980813-122980813 on Build GRCh38
rs1028935454 Chr.12: 122980812-122980812 on Build GRCh38
rs1018744243 Chr.12: 122980840-122980840 on Build GRCh38
rs935629108 Chr.12: 122980718-122980718 on Build GRCh38
rs925651028 Chr.12: 122980717-122980717 on Build GRCh38
rs930403395 Chr.12: 122980739-122980739 on Build GRCh38
rs1011537722 Chr.12: 122980797-122980797 on Build GRCh38
rs975102514 Chr.12: 122980887-122980887 on Build GRCh38
rs906671580 Chr.12: 122980760-122980760 on Build GRCh38
rs967452275 Chr.12: 122980805-122980805 on Build GRCh38
rs574604941 Chr.12: 122980919-122980919 on Build GRCh38
rs965988676 Chr.12: 122980857-122980857 on Build GRCh38
rs911784582 Chr.12: 122980716-122980716 on Build GRCh38
rs1002217306 Chr.12: 122980770-122980770 on Build GRCh38
rs562599150 Chr.12: 122980911-122980911 on Build GRCh38
rs1046625987 Chr.12: 122980750-122980750 on Build GRCh38
rs987323558 Chr.12: 122980814-122980814 on Build GRCh38
rs373271382 Chr.12: 122980800-122980800 on Build GRCh38
rs948272424 Chr.12: 122980748-122980748 on Build GRCh38
rs1048808727 Chr.12: 122980742-122980742 on Build GRCh38
rs1015383138 Chr.12: 122980771-122980771 on Build GRCh38
rs114587784 Chr.12: 122980909-122980909 on Build GRCh38
rs186157628 Chr.12: 122980890-122980890 on Build GRCh38
rs377614317 Chr.12: 122980726-122980726 on Build GRCh38
rs935757802 Chr.12: 122980885-122980885 on Build GRCh38
rs940815958 Chr.12: 122980758-122980758 on Build GRCh38
rs995280088 Chr.12: 122980801-122980801 on Build GRCh38
rs895436228 Chr.12: 122980745-122980745 on Build GRCh38
rs977884186 Chr.12: 122980807-122980807 on Build GRCh38
rs925734114 Chr.12: 122980876-122980876 on Build GRCh38
rs899244945 Chr.12: 122980785-122980785 on Build GRCh38
rs1004976175 Chr.12: 122980761-122980761 on Build GRCh38
rs977202966 Chr.12: 122980848-122980848 on Build GRCh38
rs1034350981 Chr.12: 122980784-122980784 on Build GRCh38
rs74815519 Chr.12: 122980731-122980731 on Build GRCh38

View Full Product Details