Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 119583502-119583773 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENPP2
Entrez Gene ID
5168
Gene Name
ectonucleotide pyrophosphatase/phosphodiesterase 2
Gene Aliases
ATX, ATX-X, AUTOTAXIN, LysoPLD, NPP2, PD-IALPHA, PDNP2
UniGene
Hs.190977
Gene Chromosome Location
Chr.8: 119557077-119673404 on Build GRCh38
Transcripts
NM_001040092.2 NM_001130863.2 NM_006209.4 NR_045555.1 XM_006716584.1 XM_006716585.1 XM_006716587.1 XM_017013571.1 XM_017013572.1 XM_017013573.1 XM_017013574.1 XM_017013575.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5551245 c.1697G>A p.C566Y Chr.8: 119583719-119583719 on Build GRCh38
1095465 c.1677T>C p.I559I Chr.8: 119583739-119583739 on Build GRCh38
6227908 c.1535T>C p.V512A Chr.8: 119583725-119583725 on Build GRCh38
5368513 c.1543+2T>G p.? Chr.8: 119583715-119583715 on Build GRCh38
6227907 c.1691T>C p.V564A Chr.8: 119583725-119583725 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768863918 Chr.8: 119583671-119583671 on Build GRCh38
rs759046480 Chr.8: 119583693-119583693 on Build GRCh38
rs748649436 Chr.8: 119583682-119583682 on Build GRCh38
rs145038215 Chr.8: 119583739-119583739 on Build GRCh38
rs71532464 Chr.8: 119583689-119583689 on Build GRCh38
rs770248113 Chr.8: 119583743-119583743 on Build GRCh38
rs376257645 Chr.8: 119583692-119583692 on Build GRCh38
rs200910762 Chr.8: 119583680-119583680 on Build GRCh38
rs774954814 Chr.8: 119583734-119583734 on Build GRCh38
rs984209473 Chr.8: 119583556-119583556 on Build GRCh38
rs751359427 Chr.8: 119583741-119583741 on Build GRCh38
rs958364376 Chr.8: 119583610-119583610 on Build GRCh38
rs991708002 Chr.8: 119583633-119583633 on Build GRCh38
rs182023689 Chr.8: 119583654-119583654 on Build GRCh38
rs958730067 Chr.8: 119583593-119583593 on Build GRCh38
rs543972960 Chr.8: 119583527-119583527 on Build GRCh38
rs1035324476 Chr.8: 119583578-119583578 on Build GRCh38
rs374504229 Chr.8: 119583665-119583665 on Build GRCh38
rs772692083 Chr.8: 119583690-119583690 on Build GRCh38
rs991503663 Chr.8: 119583601-119583601 on Build GRCh38
rs577451265 Chr.8: 119583663-119583663 on Build GRCh38
rs909935711 Chr.8: 119583580-119583580 on Build GRCh38
rs368406755 Chr.8: 119583740-119583740 on Build GRCh38
rs764990239 Chr.8: 119583728-119583728 on Build GRCh38

View Full Product Details