Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 95062529-95062796 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCC4
Entrez Gene ID
10257
Gene Name
ATP binding cassette subfamily C member 4
Gene Aliases
MOAT-B, MOATB, MRP4
UniGene
Hs.508423
Gene Chromosome Location
Chr.13: 95019829-95301550 on Build GRCh38
Transcripts
NM_001301829.1 NM_005845.4 XM_005254025.2 XM_017020319.1 XM_017020321.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7162769 c.3280C>G p.P1094A Chr.13: 95062790-95062790 on Build GRCh38
697192 c.3316A>G p.T1106A Chr.13: 95062754-95062754 on Build GRCh38
336029 c.3326G>C p.G1109A Chr.13: 95062744-95062744 on Build GRCh38
4881079 c.3312G>C p.L1104F Chr.13: 95062758-95062758 on Build GRCh38
5021801 c.3348G>A p.K1116K Chr.13: 95062722-95062722 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776055053 Chr.13: 95062754-95062754 on Build GRCh38
rs890487925 Chr.13: 95062570-95062570 on Build GRCh38
rs1729747 Chr.13: 95062637-95062637 on Build GRCh38
rs112360695 Chr.13: 95062605-95062606 on Build GRCh38
rs565294149 Chr.13: 95062603-95062603 on Build GRCh38
rs765938854 Chr.13: 95062745-95062745 on Build GRCh38
rs752014509 Chr.13: 95062656-95062656 on Build GRCh38
rs950505445 Chr.13: 95062668-95062668 on Build GRCh38
rs1015467315 Chr.13: 95062620-95062620 on Build GRCh38
rs781624092 Chr.13: 95062671-95062671 on Build GRCh38
rs762185878 Chr.13: 95062668-95062669 on Build GRCh38
rs745881994 Chr.13: 95062687-95062687 on Build GRCh38
rs4148538 Chr.13: 95062595-95062595 on Build GRCh38
rs199683382 Chr.13: 95062595-95062595 on Build GRCh38
rs144737820 Chr.13: 95062770-95062770 on Build GRCh38
rs769571633 Chr.13: 95062737-95062737 on Build GRCh38
rs778261640 Chr.13: 95062655-95062655 on Build GRCh38
rs763903319 Chr.13: 95062674-95062674 on Build GRCh38
rs545197651 Chr.13: 95062583-95062583 on Build GRCh38
rs774100274 Chr.13: 95062720-95062720 on Build GRCh38
rs771698317 Chr.13: 95062736-95062736 on Build GRCh38
rs937669703 Chr.13: 95062596-95062596 on Build GRCh38
rs111268334 Chr.13: 95062705-95062705 on Build GRCh38
rs765416793 Chr.13: 95062634-95062634 on Build GRCh38
rs1751034 Chr.13: 95062722-95062722 on Build GRCh38
rs11568651 Chr.13: 95062665-95062665 on Build GRCh38
rs1024388256 Chr.13: 95062708-95062708 on Build GRCh38
rs749266140 Chr.13: 95062689-95062689 on Build GRCh38
rs566618544 Chr.13: 95062716-95062716 on Build GRCh38
rs370881545 Chr.13: 95062633-95062633 on Build GRCh38
rs771919171 Chr.13: 95062558-95062558 on Build GRCh38
rs888149029 Chr.13: 95062604-95062605 on Build GRCh38
rs144222160 Chr.13: 95062740-95062740 on Build GRCh38
rs1751033 Chr.13: 95062604-95062604 on Build GRCh38
rs11568655 Chr.13: 95062760-95062760 on Build GRCh38
rs11568656 Chr.13: 95062688-95062688 on Build GRCh38

View Full Product Details