Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 34707197-34707444 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLIC6
Entrez Gene ID
54102
Gene Name
chloride intracellular channel 6
Gene Aliases
CLIC1L
UniGene
Hs.473695
Gene Chromosome Location
Chr.21: 34668953-34718227 on Build GRCh38
Transcripts
NM_001317009.1 NM_053277.2 XM_017028406.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1030451 c.1407C>T p.C469C Chr.21: 34707312-34707312 on Build GRCh38
1751623 c.1428T>C p.F476F Chr.21: 34707333-34707333 on Build GRCh38
2844305 c.1387G>A p.G463S Chr.21: 34707292-34707292 on Build GRCh38
1030453 c.1479G>A p.L493L Chr.21: 34707384-34707384 on Build GRCh38
1030452 c.1415C>A p.S472Y Chr.21: 34707320-34707320 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1042170762 Chr.21: 34707311-34707311 on Build GRCh38
rs373854435 Chr.21: 34707274-34707274 on Build GRCh38
rs375561584 Chr.21: 34707236-34707236 on Build GRCh38
rs763222719 Chr.21: 34707251-34707251 on Build GRCh38
rs1004963752 Chr.21: 34707415-34707415 on Build GRCh38
rs763976338 Chr.21: 34707381-34707381 on Build GRCh38
rs368047117 Chr.21: 34707263-34707263 on Build GRCh38
rs759463061 Chr.21: 34707264-34707264 on Build GRCh38
rs374859521 Chr.21: 34707255-34707255 on Build GRCh38
rs142788359 Chr.21: 34707355-34707355 on Build GRCh38
rs752494132 Chr.21: 34707273-34707273 on Build GRCh38
rs770187791 Chr.21: 34707241-34707241 on Build GRCh38
rs748546437 Chr.21: 34707238-34707238 on Build GRCh38
rs935659743 Chr.21: 34707322-34707322 on Build GRCh38
rs368464667 Chr.21: 34707410-34707410 on Build GRCh38
rs774443825 Chr.21: 34707350-34707350 on Build GRCh38
rs769045632 Chr.21: 34707229-34707229 on Build GRCh38
rs113064323 Chr.21: 34707277-34707277 on Build GRCh38
rs777141962 Chr.21: 34707269-34707269 on Build GRCh38
rs755661243 Chr.21: 34707347-34707347 on Build GRCh38
rs115395599 Chr.21: 34707315-34707315 on Build GRCh38
rs781413499 Chr.21: 34707402-34707403 on Build GRCh38
rs368460884 Chr.21: 34707234-34707234 on Build GRCh38
rs770130798 Chr.21: 34707325-34707325 on Build GRCh38
rs764025423 Chr.21: 34707275-34707275 on Build GRCh38
rs1034690066 Chr.21: 34707220-34707220 on Build GRCh38
rs186013554 Chr.21: 34707330-34707330 on Build GRCh38
rs747265236 Chr.21: 34707262-34707264 on Build GRCh38
rs770995089 Chr.21: 34707254-34707254 on Build GRCh38
rs773405447 Chr.21: 34707248-34707248 on Build GRCh38
rs778182329 Chr.21: 34707326-34707326 on Build GRCh38
rs868729882 Chr.21: 34707304-34707304 on Build GRCh38
rs758651883 Chr.21: 34707265-34707267 on Build GRCh38
rs369846247 Chr.21: 34707354-34707354 on Build GRCh38
rs749912710 Chr.21: 34707303-34707303 on Build GRCh38
rs375922071 Chr.21: 34707299-34707299 on Build GRCh38
rs777807885 Chr.21: 34707258-34707258 on Build GRCh38
rs764817048 Chr.21: 34707401-34707401 on Build GRCh38
rs562723940 Chr.21: 34707402-34707402 on Build GRCh38
rs753522849 Chr.21: 34707284-34707284 on Build GRCh38
rs753689366 Chr.21: 34707396-34707396 on Build GRCh38
rs138955121 Chr.21: 34707314-34707314 on Build GRCh38
rs778418723 Chr.21: 34707302-34707302 on Build GRCh38
rs760748794 Chr.21: 34707360-34707360 on Build GRCh38
rs371314860 Chr.21: 34707267-34707267 on Build GRCh38

View Full Product Details