Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 46082340-46082607 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPB2
Entrez Gene ID
1361
Gene Name
carboxypeptidase B2
Gene Aliases
CPU, PCPB, TAFI
UniGene
Hs.512937
Gene Chromosome Location
Chr.13: 46053186-46105076 on Build GRCh38
Transcripts
NM_001278541.1 NM_001872.4 XM_017020393.1 NR_046226.1 NR_046227.1

Gene Symbol
CPB2-AS1
Entrez Gene ID
100509894
Gene Name
CPB2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.626139
Gene Chromosome Location
Chr.13: 46052848-46101347 on Build GRCh38
Transcripts
NM_001278541.1 NM_001872.4 XM_017020393.1 NR_046226.1 NR_046227.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4545650 c.382G>A p.E128K Chr.13: 46082443-46082443 on Build GRCh38
3664918 c.292G>A p.V98M Chr.13: 46082533-46082533 on Build GRCh38
4168807 c.340C>T p.R114* Chr.13: 46082485-46082485 on Build GRCh38
5567315 c.374C>A p.S125* Chr.13: 46082451-46082451 on Build GRCh38
947720 c.364C>T p.Q122* Chr.13: 46082461-46082461 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148585456 Chr.13: 46082541-46082541 on Build GRCh38
rs764391460 Chr.13: 46082549-46082549 on Build GRCh38
rs374795711 Chr.13: 46082472-46082472 on Build GRCh38
rs767289329 Chr.13: 46082524-46082524 on Build GRCh38
rs201278346 Chr.13: 46082502-46082502 on Build GRCh38
rs770303867 Chr.13: 46082500-46082500 on Build GRCh38
rs766675141 Chr.13: 46082455-46082456 on Build GRCh38
rs751527920 Chr.13: 46082558-46082558 on Build GRCh38
rs751480044 Chr.13: 46082461-46082461 on Build GRCh38
rs777896037 Chr.13: 46082485-46082485 on Build GRCh38
rs757390985 Chr.13: 46082565-46082565 on Build GRCh38
rs777294025 Chr.13: 46082531-46082531 on Build GRCh38
rs962374686 Chr.13: 46082586-46082586 on Build GRCh38
rs201274702 Chr.13: 46082409-46082409 on Build GRCh38
rs763744837 Chr.13: 46082429-46082429 on Build GRCh38
rs758677294 Chr.13: 46082544-46082544 on Build GRCh38
rs114181167 Chr.13: 46082492-46082492 on Build GRCh38
rs761396353 Chr.13: 46082517-46082517 on Build GRCh38
rs369350407 Chr.13: 46082394-46082394 on Build GRCh38
rs775311912 Chr.13: 46082397-46082397 on Build GRCh38
rs1020803124 Chr.13: 46082368-46082368 on Build GRCh38
rs909189636 Chr.13: 46082404-46082404 on Build GRCh38
rs769506221 Chr.13: 46082396-46082396 on Build GRCh38
rs374137399 Chr.13: 46082501-46082501 on Build GRCh38
rs758445356 Chr.13: 46082484-46082484 on Build GRCh38
rs191429775 Chr.13: 46082471-46082471 on Build GRCh38
rs1033598032 Chr.13: 46082376-46082377 on Build GRCh38
rs760443326 Chr.13: 46082535-46082535 on Build GRCh38
rs906281434 Chr.13: 46082539-46082539 on Build GRCh38
rs533780284 Chr.13: 46082557-46082557 on Build GRCh38
rs768272393 Chr.13: 46082511-46082511 on Build GRCh38
rs375565318 Chr.13: 46082477-46082477 on Build GRCh38
rs569880920 Chr.13: 46082416-46082416 on Build GRCh38
rs189042640 Chr.13: 46082372-46082372 on Build GRCh38
rs972520459 Chr.13: 46082457-46082457 on Build GRCh38
rs763119461 Chr.13: 46082456-46082457 on Build GRCh38
rs752915693 Chr.13: 46082543-46082543 on Build GRCh38
rs746176688 Chr.13: 46082498-46082498 on Build GRCh38
rs781500861 Chr.13: 46082582-46082582 on Build GRCh38
rs769143623 Chr.13: 46082515-46082515 on Build GRCh38
rs2296642 Chr.13: 46082534-46082534 on Build GRCh38
rs202112492 Chr.13: 46082495-46082495 on Build GRCh38
rs140871119 Chr.13: 46082476-46082476 on Build GRCh38
rs747538413 Chr.13: 46082489-46082489 on Build GRCh38
rs1003467763 Chr.13: 46082374-46082374 on Build GRCh38
rs17844195 Chr.13: 46082378-46082378 on Build GRCh38
rs750124567 Chr.13: 46082474-46082474 on Build GRCh38

View Full Product Details