Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 76992079-76992328 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLN5
Entrez Gene ID
1203
Gene Name
ceroid-lipofuscinosis, neuronal 5
Gene Aliases
NCL
UniGene
Hs.30213
Gene Chromosome Location
Chr.13: 76991924-77002517 on Build GRCh38
Transcripts
NM_006493.2 XM_011534917.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4802611 c.143G>T p.S48I Chr.13: 76992094-76992094 on Build GRCh38
4147768 c.257T>G p.V86G Chr.13: 76992208-76992208 on Build GRCh38
3753744 c.234C>G p.A78A Chr.13: 76992185-76992185 on Build GRCh38
6521129 c.211G>A p.G71R Chr.13: 76992162-76992162 on Build GRCh38
4147767 c.223T>C p.W75R Chr.13: 76992174-76992174 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770903467 Chr.13: 76992151-76992151 on Build GRCh38
rs775102823 Chr.13: 76992234-76992234 on Build GRCh38
rs530381028 Chr.13: 76992131-76992131 on Build GRCh38
rs958950934 Chr.13: 76992241-76992241 on Build GRCh38
rs940819040 Chr.13: 76992261-76992261 on Build GRCh38
rs770756949 Chr.13: 76992230-76992230 on Build GRCh38
rs971404954 Chr.13: 76992191-76992191 on Build GRCh38
rs772949539 Chr.13: 76992141-76992141 on Build GRCh38
rs776490903 Chr.13: 76992231-76992231 on Build GRCh38
rs1032818633 Chr.13: 76992157-76992157 on Build GRCh38
rs386833970 Chr.13: 76992242-76992243 on Build GRCh38
rs755024034 Chr.13: 76992203-76992203 on Build GRCh38
rs757017201 Chr.13: 76992285-76992285 on Build GRCh38
rs563306322 Chr.13: 76992129-76992129 on Build GRCh38
rs376454715 Chr.13: 76992159-76992159 on Build GRCh38
rs745762281 Chr.13: 76992233-76992233 on Build GRCh38
rs781182241 Chr.13: 76992283-76992283 on Build GRCh38
rs547991726 Chr.13: 76992269-76992269 on Build GRCh38
rs1007145426 Chr.13: 76992136-76992136 on Build GRCh38
rs370320230 Chr.13: 76992218-76992218 on Build GRCh38
rs1057516814 Chr.13: 76992171-76992172 on Build GRCh38
rs112141862 Chr.13: 76992289-76992289 on Build GRCh38
rs763892391 Chr.13: 76992246-76992246 on Build GRCh38
rs371624826 Chr.13: 76992099-76992099 on Build GRCh38
rs985377780 Chr.13: 76992245-76992245 on Build GRCh38
rs199727787 Chr.13: 76992174-76992174 on Build GRCh38
rs766858440 Chr.13: 76992190-76992190 on Build GRCh38
rs780198002 Chr.13: 76992229-76992230 on Build GRCh38
rs778982551 Chr.13: 76992204-76992204 on Build GRCh38
rs373102871 Chr.13: 76992125-76992125 on Build GRCh38
rs367952803 Chr.13: 76992250-76992250 on Build GRCh38
rs760539013 Chr.13: 76992097-76992097 on Build GRCh38
rs931635211 Chr.13: 76992280-76992280 on Build GRCh38
rs541501705 Chr.13: 76992117-76992117 on Build GRCh38
rs769725211 Chr.13: 76992298-76992298 on Build GRCh38
rs753237757 Chr.13: 76992106-76992106 on Build GRCh38
rs374080049 Chr.13: 76992249-76992249 on Build GRCh38
rs754303449 Chr.13: 76992260-76992260 on Build GRCh38
rs146993892 Chr.13: 76992103-76992103 on Build GRCh38
rs1057516220 Chr.13: 76992231-76992232 on Build GRCh38
rs200348035 Chr.13: 76992182-76992182 on Build GRCh38
rs746845935 Chr.13: 76992227-76992227 on Build GRCh38
rs771855769 Chr.13: 76992140-76992140 on Build GRCh38
rs760626879 Chr.13: 76992144-76992144 on Build GRCh38
rs909356586 Chr.13: 76992251-76992251 on Build GRCh38
rs755469935 Chr.13: 76992263-76992263 on Build GRCh38
rs764790770 Chr.13: 76992175-76992175 on Build GRCh38
rs759063887 Chr.13: 76992115-76992115 on Build GRCh38
rs1057517134 Chr.13: 76992253-76992265 on Build GRCh38
rs754352180 Chr.13: 76992193-76992193 on Build GRCh38
rs768028774 Chr.13: 76992180-76992180 on Build GRCh38
rs779057868 Chr.13: 76992134-76992134 on Build GRCh38
rs104894385 Chr.13: 76992176-76992176 on Build GRCh38
rs756651200 Chr.13: 76992128-76992128 on Build GRCh38
rs138037471 Chr.13: 76992185-76992185 on Build GRCh38
rs751801723 Chr.13: 76992283-76992284 on Build GRCh38
rs752389963 Chr.13: 76992118-76992118 on Build GRCh38
rs750973360 Chr.13: 76992126-76992126 on Build GRCh38
rs780975343 Chr.13: 76992287-76992287 on Build GRCh38
rs748399349 Chr.13: 76992216-76992216 on Build GRCh38
rs757756318 Chr.13: 76992122-76992122 on Build GRCh38
rs777949085 Chr.13: 76992223-76992223 on Build GRCh38
rs9565308 Chr.13: 76992279-76992279 on Build GRCh38
rs796052344 Chr.13: 76992178-76992178 on Build GRCh38
rs796052343 Chr.13: 76992156-76992157 on Build GRCh38
rs765252923 Chr.13: 76992268-76992268 on Build GRCh38

View Full Product Details