Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 73166012-73166285 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANXA1
Entrez Gene ID
301
Gene Name
annexin A1
Gene Aliases
ANX1, LPC1
UniGene
Hs.494173
Gene Chromosome Location
Chr.9: 73151731-73170393 on Build GRCh38
Transcripts
NM_000700.2 XM_011518609.1 XM_017014657.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1462935 c.801C>T p.I267I Chr.9: 73166191-73166191 on Build GRCh38
6601288 c.739G>T p.D247Y Chr.9: 73166129-73166129 on Build GRCh38
5589673 c.765G>A p.E255E Chr.9: 73166155-73166155 on Build GRCh38
318707 c.721A>G p.T241A Chr.9: 73166111-73166111 on Build GRCh38
3907935 c.799A>G p.I267V Chr.9: 73166189-73166189 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747491856 Chr.9: 73166216-73166216 on Build GRCh38
rs752547254 Chr.9: 73166067-73166067 on Build GRCh38
rs765904278 Chr.9: 73166134-73166134 on Build GRCh38
rs762557361 Chr.9: 73166132-73166132 on Build GRCh38
rs767497482 Chr.9: 73166145-73166145 on Build GRCh38
rs148059788 Chr.9: 73166182-73166182 on Build GRCh38
rs763963884 Chr.9: 73166173-73166173 on Build GRCh38
rs775791748 Chr.9: 73166112-73166112 on Build GRCh38
rs778955601 Chr.9: 73166196-73166196 on Build GRCh38
rs370169111 Chr.9: 73166229-73166229 on Build GRCh38
rs932350680 Chr.9: 73166187-73166187 on Build GRCh38
rs772901182 Chr.9: 73166122-73166122 on Build GRCh38
rs200321397 Chr.9: 73166171-73166171 on Build GRCh38
rs915546411 Chr.9: 73166198-73166198 on Build GRCh38
rs367962910 Chr.9: 73166155-73166155 on Build GRCh38
rs756034897 Chr.9: 73166169-73166169 on Build GRCh38
rs532429182 Chr.9: 73166073-73166073 on Build GRCh38
rs146251070 Chr.9: 73166119-73166119 on Build GRCh38
rs571324236 Chr.9: 73166205-73166205 on Build GRCh38
rs751100018 Chr.9: 73166137-73166137 on Build GRCh38
rs1032094617 Chr.9: 73166254-73166254 on Build GRCh38
rs745896768 Chr.9: 73166102-73166102 on Build GRCh38
rs772657814 Chr.9: 73166109-73166109 on Build GRCh38
rs892583460 Chr.9: 73166255-73166255 on Build GRCh38
rs758401436 Chr.9: 73166208-73166208 on Build GRCh38
rs771184852 Chr.9: 73166096-73166096 on Build GRCh38
rs375801841 Chr.9: 73166058-73166058 on Build GRCh38
rs760391534 Chr.9: 73166218-73166218 on Build GRCh38
rs377359805 Chr.9: 73166115-73166115 on Build GRCh38
rs751487052 Chr.9: 73166051-73166051 on Build GRCh38
rs778210865 Chr.9: 73166080-73166080 on Build GRCh38
rs1027254126 Chr.9: 73166061-73166061 on Build GRCh38
rs112185678 Chr.9: 73166193-73166193 on Build GRCh38
rs757586479 Chr.9: 73166191-73166191 on Build GRCh38
rs568014020 Chr.9: 73166257-73166257 on Build GRCh38
rs111498712 Chr.9: 73166194-73166194 on Build GRCh38
rs143511180 Chr.9: 73166228-73166228 on Build GRCh38
rs1052548656 Chr.9: 73166247-73166247 on Build GRCh38
rs758910600 Chr.9: 73166139-73166139 on Build GRCh38
rs780668316 Chr.9: 73166212-73166212 on Build GRCh38
rs1000754947 Chr.9: 73166252-73166252 on Build GRCh38
rs959775361 Chr.9: 73166141-73166141 on Build GRCh38
rs752680857 Chr.9: 73166168-73166168 on Build GRCh38
rs973540500 Chr.9: 73166126-73166126 on Build GRCh38
rs779021366 Chr.9: 73166097-73166097 on Build GRCh38
rs764761585 Chr.9: 73166127-73166127 on Build GRCh38
rs749513198 Chr.9: 73166088-73166088 on Build GRCh38
rs780939771 Chr.9: 73166060-73166060 on Build GRCh38
rs754154590 Chr.9: 73166179-73166179 on Build GRCh38

View Full Product Details