Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1448854-1449091 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLCN7
Entrez Gene ID
1186
Gene Name
chloride voltage-gated channel 7
Gene Aliases
CLC-7, CLC7, OPTA2, OPTB4, PPP1R63
UniGene
Hs.459649
Gene Chromosome Location
Chr.16: 1444933-1475084 on Build GRCh38
Transcripts
NM_001114331.2 NM_001287.5 XM_011522354.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
137032 c.1714G>A p.E572K Chr.16: 1449049-1449049 on Build GRCh38
6143456 c.1727A>T p.N576I Chr.16: 1449036-1449036 on Build GRCh38
4987342 c.1786G>A p.V596I Chr.16: 1448977-1448977 on Build GRCh38
967301 c.1701C>T p.T567T Chr.16: 1449062-1449062 on Build GRCh38
3506149 c.1746C>T p.P582P Chr.16: 1449017-1449017 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753281228 Chr.16: 1448929-1448929 on Build GRCh38
rs752143449 Chr.16: 1449068-1449068 on Build GRCh38
rs765883547 Chr.16: 1449056-1449056 on Build GRCh38
rs779332096 Chr.16: 1448927-1448927 on Build GRCh38
rs144540326 Chr.16: 1448916-1448916 on Build GRCh38
rs762957941 Chr.16: 1448956-1448956 on Build GRCh38
rs753172781 Chr.16: 1449058-1449058 on Build GRCh38
rs758949757 Chr.16: 1448981-1448981 on Build GRCh38
rs774253483 Chr.16: 1449020-1449020 on Build GRCh38
rs200121444 Chr.16: 1448977-1448977 on Build GRCh38
rs368913050 Chr.16: 1448931-1448931 on Build GRCh38
rs201691721 Chr.16: 1449002-1449002 on Build GRCh38
rs546638957 Chr.16: 1448889-1448889 on Build GRCh38
rs760390470 Chr.16: 1449043-1449043 on Build GRCh38
rs747018809 Chr.16: 1448932-1448932 on Build GRCh38
rs966279317 Chr.16: 1448910-1448910 on Build GRCh38
rs377190713 Chr.16: 1448995-1448995 on Build GRCh38
rs766299498 Chr.16: 1448887-1448887 on Build GRCh38
rs964264070 Chr.16: 1448907-1448907 on Build GRCh38
rs528297027 Chr.16: 1448873-1448873 on Build GRCh38
rs771346530 Chr.16: 1448945-1448945 on Build GRCh38
rs768693668 Chr.16: 1449017-1449017 on Build GRCh38
rs776412250 Chr.16: 1448899-1448899 on Build GRCh38
rs1000196945 Chr.16: 1448878-1448879 on Build GRCh38
rs990837869 Chr.16: 1448908-1448908 on Build GRCh38
rs779900843 Chr.16: 1449011-1449011 on Build GRCh38
rs995877497 Chr.16: 1448912-1448912 on Build GRCh38
rs376073738 Chr.16: 1448878-1448878 on Build GRCh38
rs746112826 Chr.16: 1449000-1449000 on Build GRCh38
rs369837981 Chr.16: 1448996-1448996 on Build GRCh38
rs772664007 Chr.16: 1449035-1449035 on Build GRCh38
rs138420901 Chr.16: 1448978-1448978 on Build GRCh38
rs554219137 Chr.16: 1449061-1449061 on Build GRCh38
rs143025396 Chr.16: 1449062-1449062 on Build GRCh38
rs749027624 Chr.16: 1449013-1449013 on Build GRCh38
rs566569883 Chr.16: 1448891-1448891 on Build GRCh38
rs768408258 Chr.16: 1448958-1448958 on Build GRCh38
rs564060968 Chr.16: 1448884-1448884 on Build GRCh38
rs761356948 Chr.16: 1449026-1449026 on Build GRCh38
rs367688658 Chr.16: 1448951-1448951 on Build GRCh38
rs200682842 Chr.16: 1448962-1448962 on Build GRCh38
rs1026187628 Chr.16: 1448876-1448876 on Build GRCh38
rs755317673 Chr.16: 1448918-1448918 on Build GRCh38
rs376712645 Chr.16: 1448888-1448889 on Build GRCh38
rs113456614 Chr.16: 1448874-1448874 on Build GRCh38
rs778420950 Chr.16: 1448986-1448986 on Build GRCh38
rs994244336 Chr.16: 1448875-1448875 on Build GRCh38
rs201783415 Chr.16: 1448961-1448961 on Build GRCh38
rs760207641 Chr.16: 1448970-1448970 on Build GRCh38
rs551660579 Chr.16: 1448885-1448885 on Build GRCh38
rs751747589 Chr.16: 1448987-1448987 on Build GRCh38
rs771604430 Chr.16: 1449034-1449034 on Build GRCh38
rs12932780 Chr.16: 1448950-1448950 on Build GRCh38

View Full Product Details