Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 34874494-34874719 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AQR
Entrez Gene ID
9716
Gene Name
aquarius intron-binding spliceosomal factor
Gene Aliases
IBP160, fSAP164
UniGene
Hs.510958
Gene Chromosome Location
Chr.15: 34856351-34969794 on Build GRCh38
Transcripts
NM_014691.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7182778 c.3406C>G p.Q1136E Chr.15: 34874696-34874696 on Build GRCh38
4852076 c.3425G>A p.S1142N Chr.15: 34874677-34874677 on Build GRCh38
1245274 c.3386C>T p.P1129L Chr.15: 34874716-34874716 on Build GRCh38
5362898 c.3420A>C p.R1140S Chr.15: 34874682-34874682 on Build GRCh38
5054255 c.3404C>A p.A1135D Chr.15: 34874698-34874698 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs905142337 Chr.15: 34874598-34874598 on Build GRCh38
rs540895830 Chr.15: 34874568-34874568 on Build GRCh38
rs751301330 Chr.15: 34874586-34874586 on Build GRCh38
rs775479403 Chr.15: 34874574-34874574 on Build GRCh38
rs1009792150 Chr.15: 34874655-34874655 on Build GRCh38
rs565314234 Chr.15: 34874590-34874590 on Build GRCh38
rs376843690 Chr.15: 34874648-34874648 on Build GRCh38
rs192645883 Chr.15: 34874537-34874537 on Build GRCh38
rs778857349 Chr.15: 34874665-34874665 on Build GRCh38
rs750587400 Chr.15: 34874671-34874671 on Build GRCh38
rs922785319 Chr.15: 34874532-34874532 on Build GRCh38
rs370712067 Chr.15: 34874653-34874653 on Build GRCh38
rs1047143299 Chr.15: 34874549-34874549 on Build GRCh38
rs1039937556 Chr.15: 34874524-34874524 on Build GRCh38
rs780745443 Chr.15: 34874683-34874683 on Build GRCh38
rs184526206 Chr.15: 34874662-34874662 on Build GRCh38
rs1002073125 Chr.15: 34874618-34874618 on Build GRCh38
rs573601185 Chr.15: 34874558-34874558 on Build GRCh38
rs373849838 Chr.15: 34874663-34874663 on Build GRCh38
rs569404837 Chr.15: 34874538-34874538 on Build GRCh38
rs375574768 Chr.15: 34874636-34874636 on Build GRCh38
rs758948708 Chr.15: 34874679-34874679 on Build GRCh38
rs978081785 Chr.15: 34874652-34874652 on Build GRCh38
rs1005521880 Chr.15: 34874561-34874561 on Build GRCh38
rs756949703 Chr.15: 34874654-34874655 on Build GRCh38
rs965403789 Chr.15: 34874619-34874619 on Build GRCh38

View Full Product Details