Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 226994766-226995039 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC42BPA
Entrez Gene ID
8476
Gene Name
CDC42 binding protein kinase alpha
Gene Aliases
MRCK, MRCKA, PK428
UniGene
Hs.35433
Gene Chromosome Location
Chr.1: 226989865-227318492 on Build GRCh38
Transcripts
NM_003607.3 NM_014826.4 XM_005273317.3 XM_005273318.3 XM_005273320.3 XM_005273321.3 XM_005273322.3 XM_005273324.3 XM_006711834.3 XM_006711835.3 XM_011544306.2 XM_011544307.2 XM_011544308.2 XM_011544309.2 XM_017002578.1 XM_017002579.1 XM_017002580.1 XM_017002581.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1200362 c.4978G>A p.G1660R Chr.1: 226994873-226994873 on Build GRCh38
364239 c.4874C>T p.S1625L Chr.1: 226994977-226994977 on Build GRCh38
1339337 c.4651G>A p.A1551T Chr.1: 226994957-226994957 on Build GRCh38
363601 c.4979C>A p.S1660* Chr.1: 226994977-226994977 on Build GRCh38
5417135 c.5014T>A p.F1672I Chr.1: 226994942-226994942 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745591011 Chr.1: 226994995-226994995 on Build GRCh38
rs766359544 Chr.1: 226994829-226994829 on Build GRCh38
rs748603329 Chr.1: 226994970-226994970 on Build GRCh38
rs775288553 Chr.1: 226995002-226995002 on Build GRCh38
rs749739379 Chr.1: 226994801-226994801 on Build GRCh38
rs763735203 Chr.1: 226995014-226995014 on Build GRCh38
rs769833093 Chr.1: 226994999-226994999 on Build GRCh38
rs942325892 Chr.1: 226994987-226994987 on Build GRCh38
rs770475818 Chr.1: 226994799-226994799 on Build GRCh38
rs749751520 Chr.1: 226994873-226994873 on Build GRCh38
rs536397809 Chr.1: 226994998-226994998 on Build GRCh38
rs759429493 Chr.1: 226994926-226994926 on Build GRCh38
rs751193044 Chr.1: 226994956-226994956 on Build GRCh38
rs774733690 Chr.1: 226994892-226994892 on Build GRCh38
rs747186508 Chr.1: 226994984-226994984 on Build GRCh38
rs931142776 Chr.1: 226994854-226994854 on Build GRCh38
rs201650255 Chr.1: 226994804-226994804 on Build GRCh38
rs147810081 Chr.1: 226994958-226994958 on Build GRCh38
rs746579028 Chr.1: 226994855-226994855 on Build GRCh38
rs767520723 Chr.1: 226994810-226994810 on Build GRCh38
rs752323864 Chr.1: 226994842-226994842 on Build GRCh38
rs16846779 Chr.1: 226994973-226994973 on Build GRCh38
rs776573133 Chr.1: 226994917-226994917 on Build GRCh38
rs779265076 Chr.1: 226994964-226994964 on Build GRCh38
rs756838979 Chr.1: 226994957-226994957 on Build GRCh38
rs1044332221 Chr.1: 226994867-226994867 on Build GRCh38
rs762042217 Chr.1: 226994809-226994809 on Build GRCh38
rs372068680 Chr.1: 226994800-226994800 on Build GRCh38
rs752590269 Chr.1: 226994933-226994933 on Build GRCh38
rs758118650 Chr.1: 226994935-226994935 on Build GRCh38
rs369962125 Chr.1: 226994914-226994914 on Build GRCh38
rs777272887 Chr.1: 226994850-226994850 on Build GRCh38
rs765054481 Chr.1: 226994932-226994932 on Build GRCh38
rs201963139 Chr.1: 226994841-226994841 on Build GRCh38
rs756824948 Chr.1: 226994857-226994857 on Build GRCh38
rs754783131 Chr.1: 226994838-226994838 on Build GRCh38
rs866181712 Chr.1: 226994950-226994950 on Build GRCh38
rs145538584 Chr.1: 226994906-226994906 on Build GRCh38
rs777985904 Chr.1: 226994979-226994979 on Build GRCh38
rs200692909 Chr.1: 226994881-226994881 on Build GRCh38
rs769570260 Chr.1: 226995012-226995012 on Build GRCh38
rs755545219 Chr.1: 226994963-226994963 on Build GRCh38
rs773380856 Chr.1: 226994816-226994816 on Build GRCh38
rs1045247 Chr.1: 226994874-226994874 on Build GRCh38
rs746971062 Chr.1: 226994805-226994805 on Build GRCh38
rs746491334 Chr.1: 226994798-226994798 on Build GRCh38
rs753710619 Chr.1: 226994837-226994837 on Build GRCh38
rs748430428 Chr.1: 226994895-226994895 on Build GRCh38
rs532891873 Chr.1: 226994915-226994915 on Build GRCh38
rs780708396 Chr.1: 226994863-226994863 on Build GRCh38
rs1000205631 Chr.1: 226994789-226994789 on Build GRCh38
rs904725867 Chr.1: 226994972-226994972 on Build GRCh38
rs771388079 Chr.1: 226994973-226994974 on Build GRCh38
rs771030208 Chr.1: 226994989-226994989 on Build GRCh38
rs573592647 Chr.1: 226994847-226994847 on Build GRCh38
rs143404397 Chr.1: 226994828-226994828 on Build GRCh38
rs772385104 Chr.1: 226994897-226994897 on Build GRCh38
rs745506924 Chr.1: 226995004-226995004 on Build GRCh38
rs776732519 Chr.1: 226994992-226994992 on Build GRCh38
rs763877436 Chr.1: 226994945-226994945 on Build GRCh38
rs749841288 Chr.1: 226994960-226994960 on Build GRCh38

View Full Product Details