Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 20654154-20654427 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDOST
Entrez Gene ID
1650
Gene Name
dolichyl-diphosphooligosaccharide--protein glycosyltransferase non-catalytic subunit
Gene Aliases
AGER1, CDG1R, OKSWcl45, OST, OST48, WBP1
UniGene
Hs.523145
Gene Chromosome Location
Chr.1: 20651767-20661544 on Build GRCh38
Transcripts
NM_005216.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6305291 c.783C>A p.L261L Chr.1: 20654285-20654285 on Build GRCh38
6423650 c.820C>T p.Q274* Chr.1: 20654248-20654248 on Build GRCh38
5379521 c.800C>T p.S267F Chr.1: 20654268-20654268 on Build GRCh38
3976942 c.768C>G p.I256M Chr.1: 20654300-20654300 on Build GRCh38
182813 c.761G>A p.R254H Chr.1: 20654307-20654307 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753619089 Chr.1: 20654238-20654238 on Build GRCh38
rs768610807 Chr.1: 20654195-20654195 on Build GRCh38
rs369170920 Chr.1: 20654338-20654338 on Build GRCh38
rs138539122 Chr.1: 20654295-20654295 on Build GRCh38
rs905512927 Chr.1: 20654231-20654231 on Build GRCh38
rs867590371 Chr.1: 20654351-20654351 on Build GRCh38
rs771888017 Chr.1: 20654311-20654311 on Build GRCh38
rs961389384 Chr.1: 20654237-20654237 on Build GRCh38
rs543407020 Chr.1: 20654385-20654385 on Build GRCh38
rs984845312 Chr.1: 20654395-20654395 on Build GRCh38
rs748620550 Chr.1: 20654406-20654406 on Build GRCh38
rs34189097 Chr.1: 20654306-20654306 on Build GRCh38
rs751872569 Chr.1: 20654261-20654263 on Build GRCh38
rs138061134 Chr.1: 20654241-20654241 on Build GRCh38
rs536003962 Chr.1: 20654344-20654344 on Build GRCh38
rs607254 Chr.1: 20654387-20654387 on Build GRCh38
rs1005246235 Chr.1: 20654233-20654233 on Build GRCh38
rs1016617531 Chr.1: 20654234-20654234 on Build GRCh38
rs149114970 Chr.1: 20654285-20654285 on Build GRCh38
rs538503808 Chr.1: 20654294-20654294 on Build GRCh38
rs201591222 Chr.1: 20654384-20654384 on Build GRCh38
rs755046561 Chr.1: 20654393-20654393 on Build GRCh38
rs150551993 Chr.1: 20654305-20654305 on Build GRCh38
rs758589166 Chr.1: 20654380-20654380 on Build GRCh38
rs373387668 Chr.1: 20654321-20654321 on Build GRCh38
rs142613484 Chr.1: 20654187-20654187 on Build GRCh38
rs750286377 Chr.1: 20654363-20654363 on Build GRCh38
rs757033527 Chr.1: 20654240-20654240 on Build GRCh38
rs949892317 Chr.1: 20654358-20654358 on Build GRCh38
rs1804070 Chr.1: 20654267-20654267 on Build GRCh38
rs886045869 Chr.1: 20654234-20654234 on Build GRCh38
rs761610774 Chr.1: 20654360-20654360 on Build GRCh38
rs1044366602 Chr.1: 20654209-20654209 on Build GRCh38
rs199782279 Chr.1: 20654265-20654265 on Build GRCh38
rs780103981 Chr.1: 20654284-20654284 on Build GRCh38
rs760625768 Chr.1: 20654198-20654198 on Build GRCh38
rs755348291 Chr.1: 20654351-20654351 on Build GRCh38
rs41307775 Chr.1: 20654354-20654354 on Build GRCh38
rs955578863 Chr.1: 20654250-20654250 on Build GRCh38
rs375087961 Chr.1: 20654206-20654206 on Build GRCh38
rs775115030 Chr.1: 20654313-20654313 on Build GRCh38
rs1014175774 Chr.1: 20654202-20654202 on Build GRCh38
rs765003010 Chr.1: 20654361-20654361 on Build GRCh38
rs573385813 Chr.1: 20654357-20654357 on Build GRCh38
rs774094300 Chr.1: 20654307-20654307 on Build GRCh38
rs758677393 Chr.1: 20654272-20654272 on Build GRCh38
rs748804145 Chr.1: 20654308-20654308 on Build GRCh38
rs780866895 Chr.1: 20654249-20654249 on Build GRCh38

View Full Product Details