Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 126832472-126832745 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZBTB43
Entrez Gene ID
23099
Gene Name
zinc finger and BTB domain containing 43
Gene Aliases
ZBTB22B, ZNF-X, ZNF297B
UniGene
Hs.355581
Gene Chromosome Location
Chr.9: 126804072-126838210 on Build GRCh38
Transcripts
NM_001135776.1 NM_014007.3 XM_005251835.3 XM_011518409.1 XM_011518411.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3654383 c.47C>T p.S16F Chr.9: 126832556-126832556 on Build GRCh38
6214017 c.51C>T p.S17S Chr.9: 126832560-126832560 on Build GRCh38
4742175 c.193delA p.N66fs*10 Chr.9: 126832702-126832702 on Build GRCh38
4742176 c.221A>G p.D74G Chr.9: 126832730-126832730 on Build GRCh38
221755 c.37C>T p.P13S Chr.9: 126832546-126832546 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs578000038 Chr.9: 126832535-126832535 on Build GRCh38
rs903396679 Chr.9: 126832513-126832513 on Build GRCh38
rs113316414 Chr.9: 126832691-126832691 on Build GRCh38
rs1017295587 Chr.9: 126832635-126832635 on Build GRCh38
rs999126833 Chr.9: 126832522-126832522 on Build GRCh38
rs753157885 Chr.9: 126832686-126832686 on Build GRCh38
rs776790476 Chr.9: 126832602-126832602 on Build GRCh38
rs373620611 Chr.9: 126832642-126832642 on Build GRCh38
rs768016149 Chr.9: 126832657-126832657 on Build GRCh38
rs372479585 Chr.9: 126832524-126832524 on Build GRCh38
rs371516295 Chr.9: 126832611-126832611 on Build GRCh38
rs200997109 Chr.9: 126832656-126832656 on Build GRCh38
rs777753960 Chr.9: 126832564-126832564 on Build GRCh38
rs771054181 Chr.9: 126832596-126832596 on Build GRCh38
rs770015226 Chr.9: 126832617-126832617 on Build GRCh38
rs573779320 Chr.9: 126832643-126832643 on Build GRCh38
rs368770592 Chr.9: 126832500-126832500 on Build GRCh38
rs771744088 Chr.9: 126832537-126832537 on Build GRCh38
rs753304034 Chr.9: 126832668-126832668 on Build GRCh38
rs34789090 Chr.9: 126832641-126832641 on Build GRCh38
rs775595302 Chr.9: 126832640-126832640 on Build GRCh38
rs950657336 Chr.9: 126832713-126832713 on Build GRCh38
rs984736251 Chr.9: 126832715-126832715 on Build GRCh38
rs746942945 Chr.9: 126832578-126832578 on Build GRCh38
rs754592887 Chr.9: 126832502-126832502 on Build GRCh38

View Full Product Details