Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 32974409-32974678 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRD2
Entrez Gene ID
6046
Gene Name
bromodomain containing 2
Gene Aliases
D6S113E, FSH, FSRG1, NAT, O27.1.1, RING3, RNF3
UniGene
Hs.75243
Gene Chromosome Location
Chr.6: 32968660-32981505 on Build GRCh38
Transcripts
NM_001113182.2 NM_001199455.1 NM_001199456.1 NM_001291986.1 NM_005104.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6447337 c.242A>G p.Y81C Chr.6: 32974674-32974674 on Build GRCh38
7357720 c.68G>A p.G23D Chr.6: 32974500-32974500 on Build GRCh38
405216 c.66delG p.G23fs*63 Chr.6: 32974498-32974498 on Build GRCh38
1621497 c.39G>A p.G13G Chr.6: 32974471-32974471 on Build GRCh38
4595910 c.145G>T p.A49S Chr.6: 32974577-32974577 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200307380 Chr.6: 32974551-32974551 on Build GRCh38
rs759910342 Chr.6: 32974486-32974486 on Build GRCh38
rs763014373 Chr.6: 32974534-32974534 on Build GRCh38
rs752965005 Chr.6: 32974547-32974548 on Build GRCh38
rs1022792429 Chr.6: 32974590-32974590 on Build GRCh38
rs750228772 Chr.6: 32974522-32974522 on Build GRCh38
rs775890799 Chr.6: 32974637-32974637 on Build GRCh38
rs746604944 Chr.6: 32974443-32974443 on Build GRCh38
rs924069269 Chr.6: 32974514-32974514 on Build GRCh38
rs770137865 Chr.6: 32974581-32974581 on Build GRCh38
rs879045467 Chr.6: 32974555-32974555 on Build GRCh38
rs55650502 Chr.6: 32974500-32974500 on Build GRCh38
rs766690749 Chr.6: 32974481-32974481 on Build GRCh38
rs756964549 Chr.6: 32974507-32974507 on Build GRCh38
rs3918144 Chr.6: 32974578-32974578 on Build GRCh38
rs1035375467 Chr.6: 32974622-32974622 on Build GRCh38
rs772233506 Chr.6: 32974465-32974465 on Build GRCh38
rs35261809 Chr.6: 32974640-32974640 on Build GRCh38
rs767109720 Chr.6: 32974603-32974603 on Build GRCh38
rs773585882 Chr.6: 32974466-32974466 on Build GRCh38
rs778053233 Chr.6: 32974553-32974553 on Build GRCh38
rs748209924 Chr.6: 32974462-32974462 on Build GRCh38
rs55669504 Chr.6: 32974577-32974577 on Build GRCh38
rs371937442 Chr.6: 32974505-32974505 on Build GRCh38
rs138097363 Chr.6: 32974609-32974609 on Build GRCh38
rs757917123 Chr.6: 32974501-32974501 on Build GRCh38
rs56109539 Chr.6: 32974612-32974612 on Build GRCh38
rs776906957 Chr.6: 32974485-32974485 on Build GRCh38
rs148450256 Chr.6: 32974552-32974552 on Build GRCh38
rs376046629 Chr.6: 32974436-32974436 on Build GRCh38
rs766998848 Chr.6: 32974648-32974648 on Build GRCh38
rs747061505 Chr.6: 32974567-32974567 on Build GRCh38
rs775159619 Chr.6: 32974638-32974638 on Build GRCh38
rs753668882 Chr.6: 32974607-32974607 on Build GRCh38
rs529211144 Chr.6: 32974616-32974616 on Build GRCh38
rs375083792 Chr.6: 32974578-32974578 on Build GRCh38
rs780917555 Chr.6: 32974449-32974449 on Build GRCh38
rs902105343 Chr.6: 32974542-32974542 on Build GRCh38
rs779883033 Chr.6: 32974539-32974539 on Build GRCh38
rs774802785 Chr.6: 32974646-32974646 on Build GRCh38
rs369193752 Chr.6: 32974574-32974574 on Build GRCh38
rs3752528 Chr.6: 32974487-32974487 on Build GRCh38
rs750051591 Chr.6: 32974654-32974654 on Build GRCh38
rs516535 Chr.6: 32974525-32974525 on Build GRCh38
rs146881536 Chr.6: 32974623-32974623 on Build GRCh38
rs748849886 Chr.6: 32974473-32974473 on Build GRCh38
rs775470105 Chr.6: 32974454-32974454 on Build GRCh38
rs115120098 Chr.6: 32974633-32974633 on Build GRCh38
rs763519762 Chr.6: 32974597-32974597 on Build GRCh38
rs376374651 Chr.6: 32974639-32974639 on Build GRCh38
rs201647382 Chr.6: 32974647-32974647 on Build GRCh38
rs376209743 Chr.6: 32974548-32974548 on Build GRCh38
rs199590043 Chr.6: 32974624-32974624 on Build GRCh38
rs745511181 Chr.6: 32974558-32974558 on Build GRCh38
rs927977214 Chr.6: 32974482-32974482 on Build GRCh38
rs769905170 Chr.6: 32974635-32974635 on Build GRCh38
rs769192210 Chr.6: 32974645-32974645 on Build GRCh38
rs775865042 Chr.6: 32974596-32974596 on Build GRCh38
rs887740788 Chr.6: 32974594-32974594 on Build GRCh38
rs534193544 Chr.6: 32974447-32974447 on Build GRCh38
rs770992353 Chr.6: 32974620-32974620 on Build GRCh38
rs864309591 Chr.6: 32974467-32974467 on Build GRCh38
rs143451360 Chr.6: 32974547-32974547 on Build GRCh38
rs913961133 Chr.6: 32974493-32974493 on Build GRCh38
rs769669840 Chr.6: 32974452-32974452 on Build GRCh38
rs779835550 Chr.6: 32974605-32974605 on Build GRCh38
rs968334632 Chr.6: 32974651-32974651 on Build GRCh38
rs755878595 Chr.6: 32974604-32974604 on Build GRCh38
rs572424072 Chr.6: 32974602-32974602 on Build GRCh38
rs35974351 Chr.6: 32974465-32974465 on Build GRCh38
rs780877106 Chr.6: 32974513-32974513 on Build GRCh38
rs138578000 Chr.6: 32974446-32974447 on Build GRCh38
rs368437378 Chr.6: 32974503-32974503 on Build GRCh38

View Full Product Details