Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 70043445-70043718 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AWAT2
Entrez Gene ID
158835
Gene Name
acyl-CoA wax alcohol acyltransferase 2
Gene Aliases
ARAT, DC4, DGAT2L4, MFAT, WS
UniGene
Hs.661507
Gene Chromosome Location
Chr.X: 70040542-70049970 on Build GRCh38
Transcripts
NM_001002254.1 XM_011530876.2

SNP Detail

SNP ID Chromosome Location
rs928118155 Chr.X: 70043535-70043535 on Build GRCh38
rs939593268 Chr.X: 70043596-70043596 on Build GRCh38
rs780227458 Chr.X: 70043616-70043616 on Build GRCh38
rs61735680 Chr.X: 70043664-70043664 on Build GRCh38
rs753972356 Chr.X: 70043619-70043619 on Build GRCh38
rs370650506 Chr.X: 70043566-70043566 on Build GRCh38
rs375588673 Chr.X: 70043686-70043686 on Build GRCh38
rs770607680 Chr.X: 70043623-70043623 on Build GRCh38
rs377676561 Chr.X: 70043691-70043691 on Build GRCh38
rs897850200 Chr.X: 70043687-70043687 on Build GRCh38
rs779029391 Chr.X: 70043638-70043638 on Build GRCh38
rs774238083 Chr.X: 70043652-70043652 on Build GRCh38
rs200467732 Chr.X: 70043564-70043564 on Build GRCh38
rs148056868 Chr.X: 70043651-70043651 on Build GRCh38
rs748527493 Chr.X: 70043643-70043643 on Build GRCh38
rs61735682 Chr.X: 70043662-70043662 on Build GRCh38
rs756256604 Chr.X: 70043573-70043573 on Build GRCh38
rs757025455 Chr.X: 70043470-70043470 on Build GRCh38

View Full Product Details