Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 133479961-133480222 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOC4
Entrez Gene ID
60412
Gene Name
exocyst complex component 4
Gene Aliases
SEC8, SEC8L1, Sec8p
UniGene
Hs.321273
Gene Chromosome Location
Chr.7: 133253067-134067137 on Build GRCh38
Transcripts
NM_001037126.1 NM_021807.3 XM_005250523.4 XM_017012495.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
136632 c.1343C>T p.S448F Chr.7: 133480064-133480064 on Build GRCh38
1251587 c.1338G>A p.S446S Chr.7: 133480059-133480059 on Build GRCh38
6176062 c.1337C>T p.S446L Chr.7: 133480058-133480058 on Build GRCh38
1085717 c.1400G>A p.R467Q Chr.7: 133480121-133480121 on Build GRCh38
7108002 c.1393T>A p.Y465N Chr.7: 133480114-133480114 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775370795 Chr.7: 133480093-133480093 on Build GRCh38
rs1051139038 Chr.7: 133480176-133480176 on Build GRCh38
rs761418536 Chr.7: 133480019-133480019 on Build GRCh38
rs751704645 Chr.7: 133480191-133480191 on Build GRCh38
rs531366432 Chr.7: 133480064-133480064 on Build GRCh38
rs999893959 Chr.7: 133480046-133480046 on Build GRCh38
rs766983321 Chr.7: 133480126-133480126 on Build GRCh38
rs199516005 Chr.7: 133480067-133480067 on Build GRCh38
rs540677432 Chr.7: 133480144-133480144 on Build GRCh38
rs765608528 Chr.7: 133480157-133480157 on Build GRCh38
rs753221519 Chr.7: 133480161-133480161 on Build GRCh38
rs375773542 Chr.7: 133480012-133480012 on Build GRCh38
rs772809747 Chr.7: 133480100-133480100 on Build GRCh38
rs1021541827 Chr.7: 133480061-133480061 on Build GRCh38
rs750055029 Chr.7: 133480030-133480030 on Build GRCh38
rs984124751 Chr.7: 133480155-133480155 on Build GRCh38
rs777989073 Chr.7: 133480180-133480180 on Build GRCh38
rs1032578634 Chr.7: 133480054-133480054 on Build GRCh38
rs759044539 Chr.7: 133480174-133480174 on Build GRCh38
rs527267574 Chr.7: 133480139-133480139 on Build GRCh38
rs755803622 Chr.7: 133480031-133480031 on Build GRCh38
rs934158897 Chr.7: 133480167-133480167 on Build GRCh38
rs762788281 Chr.7: 133480006-133480006 on Build GRCh38
rs771340209 Chr.7: 133480084-133480084 on Build GRCh38
rs761705485 Chr.7: 133480121-133480121 on Build GRCh38
rs772774793 Chr.7: 133480137-133480137 on Build GRCh38
rs768440625 Chr.7: 133480116-133480116 on Build GRCh38
rs189841429 Chr.7: 133480038-133480038 on Build GRCh38
rs749115364 Chr.7: 133480115-133480115 on Build GRCh38
rs562342594 Chr.7: 133480017-133480017 on Build GRCh38
rs917088538 Chr.7: 133480187-133480187 on Build GRCh38
rs774368637 Chr.7: 133480120-133480120 on Build GRCh38
rs545066605 Chr.7: 133480058-133480058 on Build GRCh38
rs755379810 Chr.7: 133480050-133480050 on Build GRCh38
rs747233393 Chr.7: 133480083-133480083 on Build GRCh38
rs370133161 Chr.7: 133480044-133480044 on Build GRCh38
rs775495749 Chr.7: 133480001-133480001 on Build GRCh38
rs748133993 Chr.7: 133480059-133480059 on Build GRCh38
rs767108149 Chr.7: 133480028-133480028 on Build GRCh38
rs147483176 Chr.7: 133480147-133480147 on Build GRCh38
rs764108423 Chr.7: 133480007-133480007 on Build GRCh38

View Full Product Details