Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 44976299-44976526 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOSC7
Entrez Gene ID
23016
Gene Name
exosome component 7
Gene Aliases
EAP1, RRP42, Rrp42p, hRrp42p, p8
UniGene
Hs.719958
Gene Chromosome Location
Chr.3: 44975291-45012668 on Build GRCh38
Transcripts
NM_015004.3 NR_023353.1 XM_017005930.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
359792 c.24G>T p.E8D Chr.3: 44976301-44976301 on Build GRCh38
6657313 c.29_31delAGA p.K11delK Chr.3: 44976306-44976308 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs753802102 Chr.3: 44976362-44976362 on Build GRCh38
rs374024896 Chr.3: 44976339-44976339 on Build GRCh38
rs771635775 Chr.3: 44976356-44976356 on Build GRCh38
rs763715015 Chr.3: 44976361-44976361 on Build GRCh38
rs377131187 Chr.3: 44976343-44976343 on Build GRCh38
rs6441874 Chr.3: 44976413-44976413 on Build GRCh38
rs199682012 Chr.3: 44976337-44976337 on Build GRCh38
rs987020796 Chr.3: 44976393-44976393 on Build GRCh38
rs781277910 Chr.3: 44976382-44976382 on Build GRCh38
rs6441873 Chr.3: 44976412-44976412 on Build GRCh38
rs751796481 Chr.3: 44976320-44976320 on Build GRCh38
rs1052060061 Chr.3: 44976467-44976467 on Build GRCh38
rs774995664 Chr.3: 44976359-44976359 on Build GRCh38
rs149465169 Chr.3: 44976366-44976366 on Build GRCh38
rs751150159 Chr.3: 44976383-44976384 on Build GRCh38
rs753031633 Chr.3: 44976383-44976383 on Build GRCh38
rs369122051 Chr.3: 44976378-44976378 on Build GRCh38
rs989245180 Chr.3: 44976395-44976395 on Build GRCh38
rs867676316 Chr.3: 44976386-44976386 on Build GRCh38
rs755286260 Chr.3: 44976326-44976326 on Build GRCh38
rs759139058 Chr.3: 44976355-44976355 on Build GRCh38
rs749853727 Chr.3: 44976351-44976351 on Build GRCh38
rs112635883 Chr.3: 44976494-44976494 on Build GRCh38
rs564349633 Chr.3: 44976445-44976445 on Build GRCh38
rs574646729 Chr.3: 44976353-44976353 on Build GRCh38
rs557192971 Chr.3: 44976376-44976376 on Build GRCh38
rs201199988 Chr.3: 44976348-44976348 on Build GRCh38
rs750270579 Chr.3: 44976374-44976374 on Build GRCh38
rs879308556 Chr.3: 44976412-44976413 on Build GRCh38

View Full Product Details