Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 7369244-7369495 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD163L1
Entrez Gene ID
283316
Gene Name
CD163 molecule like 1
Gene Aliases
CD163B, M160, SCARI2
UniGene
Hs.631727
Gene Chromosome Location
Chr.12: 7320510-7444185 on Build GRCh38
Transcripts
NM_001297650.1 NM_174941.5 XM_011520616.1 XM_011520617.2 XM_011520618.2 XM_011520619.1 XM_011520620.1 XM_011520621.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7154157 c.3917G>C p.G1306A Chr.12: 7369479-7369479 on Build GRCh38
6553009 c.3981C>A p.A1327A Chr.12: 7369415-7369415 on Build GRCh38
5478362 c.3940C>T p.R1314W Chr.12: 7369456-7369456 on Build GRCh38
6073609 c.3920C>A p.T1307N Chr.12: 7369476-7369476 on Build GRCh38
4147453 c.4026C>T p.G1342G Chr.12: 7369370-7369370 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748649189 Chr.12: 7369279-7369279 on Build GRCh38
rs375719648 Chr.12: 7369442-7369442 on Build GRCh38
rs189017093 Chr.12: 7369277-7369277 on Build GRCh38
rs907597065 Chr.12: 7369380-7369380 on Build GRCh38
rs994608242 Chr.12: 7369430-7369430 on Build GRCh38
rs778996485 Chr.12: 7369466-7369466 on Build GRCh38
rs915091601 Chr.12: 7369289-7369289 on Build GRCh38
rs927136738 Chr.12: 7369293-7369293 on Build GRCh38
rs756556844 Chr.12: 7369455-7369455 on Build GRCh38
rs562184599 Chr.12: 7369342-7369342 on Build GRCh38
rs755916166 Chr.12: 7369299-7369299 on Build GRCh38
rs113454788 Chr.12: 7369417-7369417 on Build GRCh38
rs1042204121 Chr.12: 7369317-7369317 on Build GRCh38
rs879064908 Chr.12: 7369390-7369390 on Build GRCh38
rs757550957 Chr.12: 7369459-7369459 on Build GRCh38
rs372730324 Chr.12: 7369456-7369456 on Build GRCh38
rs1027790002 Chr.12: 7369439-7369439 on Build GRCh38
rs746003177 Chr.12: 7369473-7369473 on Build GRCh38
rs140433520 Chr.12: 7369452-7369452 on Build GRCh38
rs768198426 Chr.12: 7369437-7369437 on Build GRCh38
rs974924055 Chr.12: 7369475-7369475 on Build GRCh38
rs201057330 Chr.12: 7369418-7369418 on Build GRCh38
rs138584383 Chr.12: 7369280-7369280 on Build GRCh38
rs761477123 Chr.12: 7369379-7369379 on Build GRCh38
rs770872194 Chr.12: 7369344-7369344 on Build GRCh38
rs775389765 Chr.12: 7369370-7369370 on Build GRCh38
rs372777721 Chr.12: 7369320-7369320 on Build GRCh38
rs370119256 Chr.12: 7369445-7369445 on Build GRCh38
rs367598769 Chr.12: 7369369-7369369 on Build GRCh38
rs980378270 Chr.12: 7369288-7369288 on Build GRCh38
rs779004989 Chr.12: 7369359-7369359 on Build GRCh38
rs34264910 Chr.12: 7369359-7369360 on Build GRCh38
rs745914422 Chr.12: 7369363-7369363 on Build GRCh38
rs746815420 Chr.12: 7369375-7369375 on Build GRCh38
rs756534474 Chr.12: 7369328-7369328 on Build GRCh38
rs762550013 Chr.12: 7369424-7369424 on Build GRCh38
rs867081460 Chr.12: 7369446-7369446 on Build GRCh38
rs777875553 Chr.12: 7369337-7369337 on Build GRCh38
rs776361885 Chr.12: 7369458-7369459 on Build GRCh38
rs966303418 Chr.12: 7369471-7369471 on Build GRCh38
rs371864155 Chr.12: 7369376-7369376 on Build GRCh38

View Full Product Details