Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 31189592-31189865 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FUS
Entrez Gene ID
2521
Gene Name
FUS RNA binding protein
Gene Aliases
ALS6, ETM4, FUS1, HNRNPP2, POMP75, TLS
UniGene
Hs.46894
Gene Chromosome Location
Chr.16: 31179444-31194871 on Build GRCh38
Transcripts
NM_001170634.1 NM_001170937.1 NM_004960.3 NR_028388.2 XM_005255233.4 XM_011545781.1 XM_011545782.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7173707 c.1051A>G p.I351V Chr.16: 31189779-31189779 on Build GRCh38
6212401 c.944A>G p.K315R Chr.16: 31189672-31189672 on Build GRCh38
5715566 c.1019C>T p.S340F Chr.16: 31189747-31189747 on Build GRCh38
1252871 c.1013C>T p.T338M Chr.16: 31189741-31189741 on Build GRCh38
5806193 c.937A>G p.T313A Chr.16: 31189665-31189665 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772245489 Chr.16: 31189700-31189700 on Build GRCh38
rs779527584 Chr.16: 31189809-31189809 on Build GRCh38
rs779456444 Chr.16: 31189822-31189822 on Build GRCh38
rs775706022 Chr.16: 31189833-31189833 on Build GRCh38
rs749604252 Chr.16: 31189667-31189667 on Build GRCh38
rs200960093 Chr.16: 31189831-31189831 on Build GRCh38
rs1038385142 Chr.16: 31189628-31189628 on Build GRCh38
rs757705019 Chr.16: 31189660-31189660 on Build GRCh38
rs368342528 Chr.16: 31189636-31189636 on Build GRCh38
rs997071883 Chr.16: 31189641-31189641 on Build GRCh38
rs748632640 Chr.16: 31189640-31189640 on Build GRCh38
rs775757526 Chr.16: 31189707-31189707 on Build GRCh38
rs200156095 Chr.16: 31189837-31189837 on Build GRCh38
rs369370003 Chr.16: 31189811-31189811 on Build GRCh38
rs765049641 Chr.16: 31189806-31189806 on Build GRCh38
rs760516023 Chr.16: 31189724-31189724 on Build GRCh38
rs771216742 Chr.16: 31189679-31189679 on Build GRCh38
rs770393324 Chr.16: 31189645-31189645 on Build GRCh38
rs757864142 Chr.16: 31189631-31189631 on Build GRCh38
rs753723545 Chr.16: 31189766-31189766 on Build GRCh38
rs34752278 Chr.16: 31189699-31189700 on Build GRCh38
rs778470554 Chr.16: 31189626-31189626 on Build GRCh38
rs367842783 Chr.16: 31189808-31189808 on Build GRCh38
rs778289704 Chr.16: 31189815-31189815 on Build GRCh38
rs879237367 Chr.16: 31189830-31189830 on Build GRCh38
rs759863892 Chr.16: 31189688-31189688 on Build GRCh38
rs370262636 Chr.16: 31189629-31189629 on Build GRCh38
rs768630899 Chr.16: 31189838-31189838 on Build GRCh38
rs756855832 Chr.16: 31189622-31189622 on Build GRCh38
rs935935656 Chr.16: 31189840-31189840 on Build GRCh38
rs1025790649 Chr.16: 31189658-31189658 on Build GRCh38
rs199705472 Chr.16: 31189655-31189655 on Build GRCh38
rs374933969 Chr.16: 31189742-31189742 on Build GRCh38
rs774663066 Chr.16: 31189682-31189682 on Build GRCh38
rs761610757 Chr.16: 31189778-31189778 on Build GRCh38
rs753722607 Chr.16: 31189621-31189621 on Build GRCh38
rs111515661 Chr.16: 31189625-31189625 on Build GRCh38
rs779490965 Chr.16: 31189671-31189673 on Build GRCh38
rs200498893 Chr.16: 31189816-31189816 on Build GRCh38
rs372038288 Chr.16: 31189646-31189646 on Build GRCh38
rs754452897 Chr.16: 31189813-31189813 on Build GRCh38
rs543862509 Chr.16: 31189818-31189818 on Build GRCh38
rs955597548 Chr.16: 31189659-31189659 on Build GRCh38

View Full Product Details