Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 88149882-88150117 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C3orf38
Entrez Gene ID
285237
Gene Name
chromosome 3 open reading frame 38
Gene Aliases
-
UniGene
Hs.476944
Gene Chromosome Location
Chr.3: 88149713-88157965 on Build GRCh38
Transcripts
NM_173824.3 XM_005264745.4

SNP Detail

SNP ID Chromosome Location
rs771104001 Chr.3: 88150056-88150056 on Build GRCh38
rs374622934 Chr.3: 88150079-88150079 on Build GRCh38
rs767709909 Chr.3: 88150037-88150038 on Build GRCh38
rs545624934 Chr.3: 88149905-88149905 on Build GRCh38
rs370794446 Chr.3: 88150014-88150014 on Build GRCh38
rs867200020 Chr.3: 88150089-88150090 on Build GRCh38
rs760285512 Chr.3: 88150020-88150020 on Build GRCh38
rs760362061 Chr.3: 88150085-88150085 on Build GRCh38
rs993905014 Chr.3: 88150061-88150061 on Build GRCh38
rs934127308 Chr.3: 88150036-88150036 on Build GRCh38
rs746194084 Chr.3: 88150041-88150042 on Build GRCh38
rs755536603 Chr.3: 88150028-88150028 on Build GRCh38
rs547554987 Chr.3: 88150003-88150003 on Build GRCh38
rs758810953 Chr.3: 88150041-88150041 on Build GRCh38
rs540851058 Chr.3: 88150040-88150041 on Build GRCh38
rs776213628 Chr.3: 88150090-88150090 on Build GRCh38
rs11374933 Chr.3: 88150035-88150036 on Build GRCh38
rs890032691 Chr.3: 88150040-88150040 on Build GRCh38
rs200620200 Chr.3: 88150035-88150035 on Build GRCh38
rs962490531 Chr.3: 88149904-88149904 on Build GRCh38
rs182126923 Chr.3: 88149942-88149942 on Build GRCh38
rs552432924 Chr.3: 88150064-88150064 on Build GRCh38
rs768525776 Chr.3: 88150076-88150076 on Build GRCh38
rs765891507 Chr.3: 88150023-88150023 on Build GRCh38
rs531168408 Chr.3: 88149941-88149941 on Build GRCh38
rs763525567 Chr.3: 88150033-88150034 on Build GRCh38
rs6551277 Chr.3: 88150029-88150029 on Build GRCh38
rs768634121 Chr.3: 88150012-88150012 on Build GRCh38
rs766012998 Chr.3: 88150087-88150087 on Build GRCh38
rs746699122 Chr.3: 88150042-88150043 on Build GRCh38
rs777779273 Chr.3: 88150039-88150040 on Build GRCh38
rs756201770 Chr.3: 88150038-88150039 on Build GRCh38
rs747220013 Chr.3: 88150042-88150042 on Build GRCh38
rs80118494 Chr.3: 88150027-88150027 on Build GRCh38
rs534691098 Chr.3: 88149958-88149958 on Build GRCh38
rs762850514 Chr.3: 88150006-88150006 on Build GRCh38
rs960921070 Chr.3: 88149929-88149929 on Build GRCh38
rs771871042 Chr.3: 88150084-88150084 on Build GRCh38
rs926032154 Chr.3: 88149996-88149996 on Build GRCh38
rs112960645 Chr.3: 88149992-88149992 on Build GRCh38
rs561621456 Chr.3: 88149955-88149955 on Build GRCh38
rs369382652 Chr.3: 88149985-88149985 on Build GRCh38
rs761598057 Chr.3: 88150015-88150015 on Build GRCh38
rs374196791 Chr.3: 88150018-88150018 on Build GRCh38
rs992015138 Chr.3: 88149934-88149934 on Build GRCh38
rs760167539 Chr.3: 88150032-88150033 on Build GRCh38
rs566096801 Chr.3: 88149924-88149924 on Build GRCh38
rs761558941 Chr.3: 88150083-88150083 on Build GRCh38
rs898248459 Chr.3: 88150052-88150052 on Build GRCh38
rs753801366 Chr.3: 88150034-88150035 on Build GRCh38

View Full Product Details