Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 3728024-3728273 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CREBBP
Entrez Gene ID
1387
Gene Name
CREB binding protein
Gene Aliases
CBP, KAT3A, RSTS
UniGene
Hs.459759
Gene Chromosome Location
Chr.16: 3725054-3880727 on Build GRCh38
Transcripts
NM_001079846.1 NM_004380.2 XM_005255124.4 XM_005255125.4 XM_006720848.3 XM_011522381.2 XM_017022944.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6492954 c.6825_6826insG p.Q2276fs*65 Chr.16: 3728221-3728222 on Build GRCh38
6016024 c.7013C>G p.S2338C Chr.16: 3728034-3728034 on Build GRCh38
254631 c.6983C>T p.S2328L Chr.16: 3728064-3728064 on Build GRCh38
3509550 c.7005C>T p.I2335I Chr.16: 3728042-3728042 on Build GRCh38
6635346 c.6846C>T p.D2282D Chr.16: 3728201-3728201 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749660119 Chr.16: 3728217-3728217 on Build GRCh38
rs770230157 Chr.16: 3728063-3728063 on Build GRCh38
rs761951767 Chr.16: 3728235-3728235 on Build GRCh38
rs996280072 Chr.16: 3728075-3728075 on Build GRCh38
rs750251074 Chr.16: 3728162-3728162 on Build GRCh38
rs753192214 Chr.16: 3728209-3728209 on Build GRCh38
rs949743163 Chr.16: 3728202-3728202 on Build GRCh38
rs979662378 Chr.16: 3728107-3728107 on Build GRCh38
rs148904096 Chr.16: 3728198-3728198 on Build GRCh38
rs745533052 Chr.16: 3728078-3728078 on Build GRCh38
rs149723395 Chr.16: 3728114-3728114 on Build GRCh38
rs771682806 Chr.16: 3728087-3728087 on Build GRCh38
rs760140366 Chr.16: 3728094-3728094 on Build GRCh38
rs771721926 Chr.16: 3728218-3728218 on Build GRCh38
rs776123525 Chr.16: 3728251-3728251 on Build GRCh38
rs372866305 Chr.16: 3728213-3728213 on Build GRCh38
rs781438241 Chr.16: 3728208-3728208 on Build GRCh38
rs761751428 Chr.16: 3728115-3728115 on Build GRCh38
rs201161108 Chr.16: 3728195-3728195 on Build GRCh38
rs1024628646 Chr.16: 3728067-3728067 on Build GRCh38
rs587778215 Chr.16: 3728091-3728091 on Build GRCh38
rs763223282 Chr.16: 3728167-3728167 on Build GRCh38
rs956984557 Chr.16: 3728159-3728159 on Build GRCh38
rs867563115 Chr.16: 3728140-3728140 on Build GRCh38
rs779528907 Chr.16: 3728234-3728234 on Build GRCh38
rs748072121 Chr.16: 3728252-3728252 on Build GRCh38
rs368174171 Chr.16: 3728194-3728194 on Build GRCh38
rs201956327 Chr.16: 3728253-3728253 on Build GRCh38
rs759722245 Chr.16: 3728223-3728240 on Build GRCh38
rs866381658 Chr.16: 3728206-3728206 on Build GRCh38
rs751654145 Chr.16: 3728141-3728142 on Build GRCh38
rs748688552 Chr.16: 3728061-3728061 on Build GRCh38
rs768095952 Chr.16: 3728246-3728246 on Build GRCh38
rs778189658 Chr.16: 3728216-3728216 on Build GRCh38
rs968610522 Chr.16: 3728080-3728080 on Build GRCh38
rs746692385 Chr.16: 3728244-3728244 on Build GRCh38
rs768181832 Chr.16: 3728106-3728106 on Build GRCh38
rs377018267 Chr.16: 3728050-3728050 on Build GRCh38
rs764944681 Chr.16: 3728117-3728117 on Build GRCh38
rs145551451 Chr.16: 3728120-3728120 on Build GRCh38

View Full Product Details