Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 81751380-81751653 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM46A
Entrez Gene ID
55603
Gene Name
family with sequence similarity 46 member A
Gene Aliases
C6orf37, XTP11
UniGene
Hs.10784
Gene Chromosome Location
Chr.6: 81745730-81752711 on Build GRCh38
Transcripts
NM_017633.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4972470 c.511G>C p.E171Q Chr.6: 81751631-81751631 on Build GRCh38
4948549 c.766A>G p.K256E Chr.6: 81751619-81751619 on Build GRCh38
5943818 c.771_772delGA p.K258fs*16 Chr.6: 81751613-81751614 on Build GRCh38
4682989 c.550A>T p.K184* Chr.6: 81751592-81751592 on Build GRCh38
4388174 c.737T>A p.L246Q Chr.6: 81751648-81751648 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778986460 Chr.6: 81751571-81751571 on Build GRCh38
rs577463790 Chr.6: 81751529-81751529 on Build GRCh38
rs1025788896 Chr.6: 81751581-81751581 on Build GRCh38
rs748052539 Chr.6: 81751624-81751624 on Build GRCh38
rs942720791 Chr.6: 81751411-81751411 on Build GRCh38
rs772776576 Chr.6: 81751632-81751632 on Build GRCh38
rs780995645 Chr.6: 81751586-81751586 on Build GRCh38
rs999871233 Chr.6: 81751503-81751503 on Build GRCh38
rs1039609846 Chr.6: 81751440-81751440 on Build GRCh38
rs182392570 Chr.6: 81751465-81751465 on Build GRCh38
rs1054629957 Chr.6: 81751519-81751519 on Build GRCh38
rs1041492021 Chr.6: 81751470-81751470 on Build GRCh38
rs752852977 Chr.6: 81751474-81751474 on Build GRCh38
rs756638909 Chr.6: 81751551-81751551 on Build GRCh38
rs548936450 Chr.6: 81751438-81751438 on Build GRCh38
rs903030676 Chr.6: 81751491-81751491 on Build GRCh38
rs753174425 Chr.6: 81751540-81751540 on Build GRCh38
rs909861857 Chr.6: 81751401-81751401 on Build GRCh38
rs185086412 Chr.6: 81751565-81751565 on Build GRCh38
rs149537489 Chr.6: 81751520-81751520 on Build GRCh38
rs773099311 Chr.6: 81751535-81751535 on Build GRCh38
rs201145878 Chr.6: 81751552-81751552 on Build GRCh38
rs749310384 Chr.6: 81751554-81751554 on Build GRCh38
rs75207242 Chr.6: 81751579-81751579 on Build GRCh38
rs537245409 Chr.6: 81751532-81751532 on Build GRCh38
rs769751143 Chr.6: 81751417-81751417 on Build GRCh38
rs970931035 Chr.6: 81751545-81751545 on Build GRCh38
rs9341898 Chr.6: 81751449-81751449 on Build GRCh38
rs761736382 Chr.6: 81751610-81751612 on Build GRCh38
rs746864011 Chr.6: 81751574-81751574 on Build GRCh38
rs919175113 Chr.6: 81751506-81751506 on Build GRCh38
rs769653047 Chr.6: 81751625-81751625 on Build GRCh38

View Full Product Details