Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 176324592-176324865 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SIMC1
Entrez Gene ID
375484
Gene Name
SUMO interacting motifs containing 1
Gene Aliases
C5orf25, OOMA1, PLEIAD
UniGene
Hs.719847
Gene Chromosome Location
Chr.5: 176238359-176345991 on Build GRCh38
Transcripts
NM_001308195.1 NM_001308196.1 NM_001308200.1 NM_198567.5 NR_131772.1 XM_011534553.2 XM_011534554.2 XM_011534556.2 XM_017009454.1 XM_017009455.1 XM_017009456.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7205285 c.411G>T p.E137D Chr.5: 176324671-176324671 on Build GRCh38
6103356 c.495G>C p.Q165H Chr.5: 176324755-176324755 on Build GRCh38
7266640 c.2023G>A p.V675I Chr.5: 176324666-176324666 on Build GRCh38
3827718 c.413T>G p.V138G Chr.5: 176324673-176324673 on Build GRCh38
7266641 c.406G>A p.V136I Chr.5: 176324666-176324666 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs919501376 Chr.5: 176324781-176324781 on Build GRCh38
rs144888692 Chr.5: 176324710-176324710 on Build GRCh38
rs909348173 Chr.5: 176324799-176324799 on Build GRCh38
rs774123626 Chr.5: 176324626-176324626 on Build GRCh38
rs771885826 Chr.5: 176324715-176324715 on Build GRCh38
rs923224799 Chr.5: 176324778-176324778 on Build GRCh38
rs750776958 Chr.5: 176324792-176324792 on Build GRCh38
rs770464365 Chr.5: 176324621-176324621 on Build GRCh38
rs771556150 Chr.5: 176324631-176324631 on Build GRCh38
rs768684470 Chr.5: 176324723-176324723 on Build GRCh38
rs760650672 Chr.5: 176324650-176324650 on Build GRCh38
rs551613175 Chr.5: 176324659-176324659 on Build GRCh38
rs779298779 Chr.5: 176324791-176324791 on Build GRCh38
rs910349709 Chr.5: 176324741-176324741 on Build GRCh38
rs370507574 Chr.5: 176324762-176324762 on Build GRCh38
rs191300514 Chr.5: 176324683-176324683 on Build GRCh38
rs905320090 Chr.5: 176324804-176324804 on Build GRCh38
rs746569008 Chr.5: 176324717-176324717 on Build GRCh38
rs140278139 Chr.5: 176324700-176324700 on Build GRCh38
rs778069897 Chr.5: 176324616-176324616 on Build GRCh38
rs374180545 Chr.5: 176324774-176324774 on Build GRCh38
rs780533741 Chr.5: 176324803-176324804 on Build GRCh38
rs777929758 Chr.5: 176324740-176324740 on Build GRCh38
rs1029513719 Chr.5: 176324660-176324660 on Build GRCh38
rs751865201 Chr.5: 176324673-176324673 on Build GRCh38
rs759762997 Chr.5: 176324738-176324738 on Build GRCh38
rs752802990 Chr.5: 176324685-176324685 on Build GRCh38
rs776545672 Chr.5: 176324664-176324664 on Build GRCh38
rs200063925 Chr.5: 176324614-176324614 on Build GRCh38
rs757711845 Chr.5: 176324784-176324784 on Build GRCh38
rs763993420 Chr.5: 176324769-176324769 on Build GRCh38
rs566159528 Chr.5: 176324716-176324716 on Build GRCh38
rs767729298 Chr.5: 176324795-176324803 on Build GRCh38
rs116614559 Chr.5: 176324641-176324641 on Build GRCh38
rs750599175 Chr.5: 176324800-176324803 on Build GRCh38
rs749681123 Chr.5: 176324618-176324618 on Build GRCh38
rs146617615 Chr.5: 176324666-176324666 on Build GRCh38
rs142932031 Chr.5: 176324792-176324793 on Build GRCh38
rs1002378226 Chr.5: 176324809-176324809 on Build GRCh38
rs756562509 Chr.5: 176324693-176324693 on Build GRCh38
rs776699051 Chr.5: 176324731-176324731 on Build GRCh38
rs374216303 Chr.5: 176324734-176324734 on Build GRCh38
rs758101800 Chr.5: 176324793-176324793 on Build GRCh38
rs764574674 Chr.5: 176324783-176324783 on Build GRCh38
rs1049343602 Chr.5: 176324797-176324797 on Build GRCh38
rs756275925 Chr.5: 176324804-176324807 on Build GRCh38
rs112164075 Chr.5: 176324818-176324818 on Build GRCh38
rs201617847 Chr.5: 176324803-176324803 on Build GRCh38
rs557440428 Chr.5: 176324820-176324820 on Build GRCh38
rs745461076 Chr.5: 176324711-176324711 on Build GRCh38
rs766505460 Chr.5: 176324672-176324672 on Build GRCh38
rs116389153 Chr.5: 176324674-176324674 on Build GRCh38
rs150336230 Chr.5: 176324707-176324707 on Build GRCh38
rs761837422 Chr.5: 176324793-176324793 on Build GRCh38
rs201370648 Chr.5: 176324732-176324732 on Build GRCh38
rs553209668 Chr.5: 176324736-176324736 on Build GRCh38

View Full Product Details