Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 53238348-53238599 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF677
Entrez Gene ID
342926
Gene Name
zinc finger protein 677
Gene Aliases
-
UniGene
Hs.20506
Gene Chromosome Location
Chr.19: 53235381-53254903 on Build GRCh38
Transcripts
NM_001317998.1 NM_182609.3 XM_005258862.4 XM_011526912.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
297651 c.214A>C p.N72H Chr.19: 53238513-53238513 on Build GRCh38
1612732 c.356G>A p.G119E Chr.19: 53238371-53238371 on Build GRCh38
1612733 c.204A>G p.S68S Chr.19: 53238523-53238523 on Build GRCh38
7124657 c.366G>C p.L122F Chr.19: 53238361-53238361 on Build GRCh38
3981708 c.257A>C p.K86T Chr.19: 53238470-53238470 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376951290 Chr.19: 53238557-53238557 on Build GRCh38
rs761505018 Chr.19: 53238458-53238458 on Build GRCh38
rs980735560 Chr.19: 53238399-53238399 on Build GRCh38
rs556293671 Chr.19: 53238484-53238484 on Build GRCh38
rs149632331 Chr.19: 53238554-53238554 on Build GRCh38
rs1010488502 Chr.19: 53238555-53238555 on Build GRCh38
rs936644255 Chr.19: 53238483-53238483 on Build GRCh38
rs921037994 Chr.19: 53238512-53238512 on Build GRCh38
rs200805678 Chr.19: 53238570-53238570 on Build GRCh38
rs768213786 Chr.19: 53238381-53238381 on Build GRCh38
rs772851234 Chr.19: 53238532-53238532 on Build GRCh38
rs746757627 Chr.19: 53238378-53238378 on Build GRCh38
rs760114973 Chr.19: 53238424-53238424 on Build GRCh38
rs772280053 Chr.19: 53238569-53238570 on Build GRCh38
rs764255088 Chr.19: 53238449-53238449 on Build GRCh38
rs747788663 Chr.19: 53238520-53238520 on Build GRCh38
rs754070234 Chr.19: 53238558-53238558 on Build GRCh38
rs373772337 Chr.19: 53238524-53238524 on Build GRCh38
rs201826579 Chr.19: 53238433-53238433 on Build GRCh38
rs775055816 Chr.19: 53238419-53238419 on Build GRCh38
rs140248775 Chr.19: 53238402-53238402 on Build GRCh38
rs748388031 Chr.19: 53238531-53238532 on Build GRCh38
rs888106858 Chr.19: 53238559-53238559 on Build GRCh38
rs780753676 Chr.19: 53238502-53238502 on Build GRCh38
rs137958797 Chr.19: 53238468-53238468 on Build GRCh38
rs772730456 Chr.19: 53238548-53238548 on Build GRCh38
rs368440040 Chr.19: 53238531-53238531 on Build GRCh38
rs764818106 Chr.19: 53238417-53238417 on Build GRCh38
rs750429688 Chr.19: 53238480-53238480 on Build GRCh38
rs371425174 Chr.19: 53238496-53238496 on Build GRCh38
rs754796600 Chr.19: 53238374-53238379 on Build GRCh38
rs866250234 Chr.19: 53238516-53238516 on Build GRCh38
rs145342344 Chr.19: 53238414-53238414 on Build GRCh38
rs746809439 Chr.19: 53238492-53238492 on Build GRCh38
rs1042399123 Chr.19: 53238566-53238566 on Build GRCh38
rs1139315 Chr.19: 53238382-53238382 on Build GRCh38
rs535354828 Chr.19: 53238469-53238469 on Build GRCh38
rs912232846 Chr.19: 53238472-53238472 on Build GRCh38
rs375245682 Chr.19: 53238396-53238396 on Build GRCh38
rs768069695 Chr.19: 53238426-53238426 on Build GRCh38
rs75924222 Chr.19: 53238491-53238491 on Build GRCh38
rs138282537 Chr.19: 53238552-53238552 on Build GRCh38
rs753018763 Chr.19: 53238428-53238428 on Build GRCh38
rs191125688 Chr.19: 53238536-53238536 on Build GRCh38
rs757381090 Chr.19: 53238461-53238461 on Build GRCh38
rs138519704 Chr.19: 53238374-53238374 on Build GRCh38
rs529539649 Chr.19: 53238379-53238380 on Build GRCh38
rs564138989 Chr.19: 53238377-53238377 on Build GRCh38
rs200224502 Chr.19: 53238487-53238487 on Build GRCh38
rs779347104 Chr.19: 53238469-53238472 on Build GRCh38

View Full Product Details