Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 61119541-61119772 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD5
Entrez Gene ID
921
Gene Name
CD5 molecule
Gene Aliases
LEU1, T1
UniGene
Hs.58685
Gene Chromosome Location
Chr.11: 61102458-61127852 on Build GRCh38
Transcripts
NM_014207.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6014449 c.774A>C p.K258N Chr.11: 61119544-61119544 on Build GRCh38
4402784 c.797T>C p.L266P Chr.11: 61119567-61119567 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs887152273 Chr.11: 61119656-61119656 on Build GRCh38
rs775270359 Chr.11: 61119574-61119574 on Build GRCh38
rs200152277 Chr.11: 61119563-61119563 on Build GRCh38
rs771219831 Chr.11: 61119562-61119562 on Build GRCh38
rs1026230106 Chr.11: 61119630-61119630 on Build GRCh38
rs759533294 Chr.11: 61119571-61119571 on Build GRCh38
rs749597421 Chr.11: 61119624-61119624 on Build GRCh38
rs749836069 Chr.11: 61119723-61119723 on Build GRCh38
rs749948985 Chr.11: 61119605-61119605 on Build GRCh38
rs375754861 Chr.11: 61119659-61119659 on Build GRCh38
rs756640975 Chr.11: 61119620-61119620 on Build GRCh38
rs1035203742 Chr.11: 61119708-61119708 on Build GRCh38
rs573854009 Chr.11: 61119657-61119657 on Build GRCh38
rs757832365 Chr.11: 61119615-61119615 on Build GRCh38
rs755572990 Chr.11: 61119610-61119610 on Build GRCh38
rs756931333 Chr.11: 61119579-61119579 on Build GRCh38
rs901357716 Chr.11: 61119603-61119603 on Build GRCh38
rs1002396990 Chr.11: 61119670-61119670 on Build GRCh38
rs533565775 Chr.11: 61119578-61119578 on Build GRCh38
rs569763809 Chr.11: 61119688-61119688 on Build GRCh38
rs887389414 Chr.11: 61119636-61119636 on Build GRCh38
rs764803429 Chr.11: 61119595-61119595 on Build GRCh38
rs780571120 Chr.11: 61119661-61119661 on Build GRCh38
rs79295876 Chr.11: 61119586-61119586 on Build GRCh38
rs371383970 Chr.11: 61119622-61119622 on Build GRCh38
rs772108327 Chr.11: 61119573-61119573 on Build GRCh38
rs753178096 Chr.11: 61119618-61119618 on Build GRCh38
rs553992183 Chr.11: 61119581-61119581 on Build GRCh38
rs1016957698 Chr.11: 61119683-61119683 on Build GRCh38

View Full Product Details