Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 122362863-122363136 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC58
Entrez Gene ID
131076
Gene Name
coiled-coil domain containing 58
Gene Aliases
-
UniGene
Hs.220594
Gene Chromosome Location
Chr.3: 122359587-122383281 on Build GRCh38
Transcripts
NM_001017928.3 NM_001308326.1 XM_011512410.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3121363 c.370G>C p.D124H Chr.3: 122362982-122362982 on Build GRCh38
6511437 c.363G>T p.V121V Chr.3: 122362989-122362989 on Build GRCh38
5764500 c.325-4G>T p.? Chr.3: 122363031-122363031 on Build GRCh38
1670510 c.343G>T p.E115* Chr.3: 122363009-122363009 on Build GRCh38
5763109 c.325-7C>T p.? Chr.3: 122363034-122363034 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1023782058 Chr.3: 122362928-122362928 on Build GRCh38
rs955174578 Chr.3: 122362895-122362895 on Build GRCh38
rs72972760 Chr.3: 122363038-122363038 on Build GRCh38
rs200634146 Chr.3: 122362951-122362951 on Build GRCh38
rs760955558 Chr.3: 122362937-122362937 on Build GRCh38
rs780803764 Chr.3: 122363073-122363073 on Build GRCh38
rs753109777 Chr.3: 122363035-122363035 on Build GRCh38
rs899624064 Chr.3: 122363094-122363094 on Build GRCh38
rs747300141 Chr.3: 122363032-122363032 on Build GRCh38
rs758743908 Chr.3: 122363016-122363016 on Build GRCh38
rs914918272 Chr.3: 122363055-122363055 on Build GRCh38
rs771642830 Chr.3: 122362980-122362980 on Build GRCh38
rs778208140 Chr.3: 122363023-122363023 on Build GRCh38
rs763936746 Chr.3: 122362925-122362925 on Build GRCh38
rs987747842 Chr.3: 122363046-122363046 on Build GRCh38
rs779599766 Chr.3: 122362984-122362984 on Build GRCh38
rs762724680 Chr.3: 122362923-122362923 on Build GRCh38
rs745370806 Chr.3: 122363073-122363073 on Build GRCh38
rs963482644 Chr.3: 122362902-122362902 on Build GRCh38
rs748845494 Chr.3: 122363038-122363072 on Build GRCh38
rs756807817 Chr.3: 122363047-122363081 on Build GRCh38
rs753986776 Chr.3: 122362967-122362967 on Build GRCh38
rs778222092 Chr.3: 122363047-122363073 on Build GRCh38
rs199808163 Chr.3: 122362979-122362979 on Build GRCh38
rs752109851 Chr.3: 122362952-122362954 on Build GRCh38
rs865801597 Chr.3: 122362963-122362963 on Build GRCh38
rs547949134 Chr.3: 122362898-122362898 on Build GRCh38
rs915216887 Chr.3: 122363066-122363066 on Build GRCh38
rs182651357 Chr.3: 122363078-122363078 on Build GRCh38
rs777514165 Chr.3: 122363035-122363042 on Build GRCh38
rs986690331 Chr.3: 122362903-122362903 on Build GRCh38
rs374071369 Chr.3: 122363052-122363052 on Build GRCh38
rs376971421 Chr.3: 122363098-122363098 on Build GRCh38
rs755218582 Chr.3: 122363034-122363035 on Build GRCh38
rs763582829 Chr.3: 122362944-122362944 on Build GRCh38
rs973910458 Chr.3: 122362905-122362905 on Build GRCh38
rs533665722 Chr.3: 122363006-122363006 on Build GRCh38
rs751254058 Chr.3: 122362931-122362931 on Build GRCh38
rs756440085 Chr.3: 122363035-122363036 on Build GRCh38
rs1028106929 Chr.3: 122362891-122362891 on Build GRCh38
rs1004886659 Chr.3: 122362896-122362896 on Build GRCh38

View Full Product Details