Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 78057957-78058194 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRAS1
Entrez Gene ID
80144
Gene Name
Fraser extracellular matrix complex subunit 1
Gene Aliases
-
UniGene
Hs.369448
Gene Chromosome Location
Chr.4: 78057323-78544269 on Build GRCh38
Transcripts
NM_001166133.1 NM_025074.6 XM_006714316.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5866768 c.60delG p.L20fs*59 Chr.4: 78058069-78058069 on Build GRCh38
7254466 c.70_71insTTCTC p.S24fs*57 Chr.4: 78058079-78058080 on Build GRCh38
5866770 c.60delG p.L20fs*59 Chr.4: 78058069-78058069 on Build GRCh38
3661199 c.62C>A p.P21H Chr.4: 78058071-78058071 on Build GRCh38
7254465 c.70_71insTTCTC p.S24fs*57 Chr.4: 78058079-78058080 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs71214392 Chr.4: 78058120-78058121 on Build GRCh38
rs775418796 Chr.4: 78058102-78058119 on Build GRCh38
rs886059625 Chr.4: 78058090-78058090 on Build GRCh38
rs768691268 Chr.4: 78058009-78058009 on Build GRCh38
rs772512930 Chr.4: 78058034-78058034 on Build GRCh38
rs111997642 Chr.4: 78058122-78058122 on Build GRCh38
rs753332895 Chr.4: 78058128-78058133 on Build GRCh38
rs771277097 Chr.4: 78058116-78058121 on Build GRCh38
rs1031980282 Chr.4: 78058146-78058146 on Build GRCh38
rs957720711 Chr.4: 78058153-78058153 on Build GRCh38
rs529437383 Chr.4: 78058121-78058121 on Build GRCh38
rs776320503 Chr.4: 78058075-78058075 on Build GRCh38
rs757054268 Chr.4: 78058109-78058109 on Build GRCh38
rs748642523 Chr.4: 78058021-78058021 on Build GRCh38
rs764024963 Chr.4: 78058104-78058104 on Build GRCh38
rs763362575 Chr.4: 78058106-78058107 on Build GRCh38
rs775323324 Chr.4: 78058128-78058129 on Build GRCh38
rs751341719 Chr.4: 78058105-78058105 on Build GRCh38
rs751308534 Chr.4: 78057994-78057994 on Build GRCh38
rs759154940 Chr.4: 78058081-78058081 on Build GRCh38
rs773457837 Chr.4: 78058048-78058048 on Build GRCh38
rs765415451 Chr.4: 78057991-78057991 on Build GRCh38
rs867934126 Chr.4: 78058079-78058079 on Build GRCh38
rs747379706 Chr.4: 78058129-78058129 on Build GRCh38
rs34237418 Chr.4: 78057987-78057987 on Build GRCh38
rs374683314 Chr.4: 78058132-78058132 on Build GRCh38
rs760594830 Chr.4: 78058127-78058128 on Build GRCh38
rs755111137 Chr.4: 78058117-78058117 on Build GRCh38
rs71661147 Chr.4: 78058103-78058104 on Build GRCh38
rs887787419 Chr.4: 78058073-78058073 on Build GRCh38
rs751915838 Chr.4: 78058101-78058102 on Build GRCh38
rs374397187 Chr.4: 78058063-78058063 on Build GRCh38
rs749377838 Chr.4: 78058005-78058005 on Build GRCh38
rs771852813 Chr.4: 78058119-78058120 on Build GRCh38
rs778297886 Chr.4: 78058125-78058125 on Build GRCh38
rs1036618746 Chr.4: 78058103-78058103 on Build GRCh38
rs367976690 Chr.4: 78058115-78058115 on Build GRCh38
rs897961753 Chr.4: 78058119-78058119 on Build GRCh38
rs762995298 Chr.4: 78058101-78058101 on Build GRCh38
rs554995505 Chr.4: 78058056-78058056 on Build GRCh38
rs780282144 Chr.4: 78058001-78058001 on Build GRCh38
rs574924373 Chr.4: 78058082-78058082 on Build GRCh38
rs752648327 Chr.4: 78057989-78057989 on Build GRCh38
rs750662462 Chr.4: 78057992-78057992 on Build GRCh38
rs764832667 Chr.4: 78058083-78058083 on Build GRCh38
rs778981554 Chr.4: 78058118-78058118 on Build GRCh38
rs794726952 Chr.4: 78058080-78058080 on Build GRCh38
rs372008113 Chr.4: 78058000-78058000 on Build GRCh38
rs201628738 Chr.4: 78058069-78058069 on Build GRCh38
rs766550602 Chr.4: 78058110-78058110 on Build GRCh38
rs962463875 Chr.4: 78058138-78058138 on Build GRCh38
rs754057254 Chr.4: 78058114-78058114 on Build GRCh38
rs774896495 Chr.4: 78058105-78058106 on Build GRCh38
rs144106232 Chr.4: 78058104-78058105 on Build GRCh38
rs774332139 Chr.4: 78058115-78058116 on Build GRCh38
rs775624675 Chr.4: 78058087-78058087 on Build GRCh38
rs796475654 Chr.4: 78058120-78058121 on Build GRCh38

View Full Product Details