Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10869159-10869432 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C19orf38
Entrez Gene ID
255809
Gene Name
chromosome 19 open reading frame 38
Gene Aliases
HIDE1
UniGene
Hs.440535
Gene Chromosome Location
Chr.19: 10836572-10869791 on Build GRCh38
Transcripts
NM_001136482.1 XM_005259846.4 XM_005259847.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1589639 c.687C>T p.C229C Chr.19: 10869361-10869361 on Build GRCh38
5732373 c.548C>T p.T183M Chr.19: 10869222-10869222 on Build GRCh38
7358173 c.592A>G p.T198A Chr.19: 10869266-10869266 on Build GRCh38
3770503 c.570C>T p.A190A Chr.19: 10869244-10869244 on Build GRCh38
1136095 c.549G>A p.T183T Chr.19: 10869223-10869223 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751901147 Chr.19: 10869278-10869278 on Build GRCh38
rs558824793 Chr.19: 10869399-10869399 on Build GRCh38
rs1048670411 Chr.19: 10869338-10869338 on Build GRCh38
rs1055314739 Chr.19: 10869383-10869383 on Build GRCh38
rs774673627 Chr.19: 10869297-10869297 on Build GRCh38
rs900490201 Chr.19: 10869194-10869194 on Build GRCh38
rs1049133624 Chr.19: 10869248-10869248 on Build GRCh38
rs969716991 Chr.19: 10869354-10869354 on Build GRCh38
rs780437963 Chr.19: 10869276-10869276 on Build GRCh38
rs911725355 Chr.19: 10869411-10869411 on Build GRCh38
rs536873152 Chr.19: 10869228-10869228 on Build GRCh38
rs933676170 Chr.19: 10869247-10869247 on Build GRCh38
rs766689265 Chr.19: 10869185-10869185 on Build GRCh38
rs771244198 Chr.19: 10869385-10869385 on Build GRCh38
rs985971411 Chr.19: 10869254-10869255 on Build GRCh38
rs930178239 Chr.19: 10869337-10869337 on Build GRCh38
rs769708747 Chr.19: 10869324-10869324 on Build GRCh38
rs748268032 Chr.19: 10869316-10869316 on Build GRCh38
rs886623044 Chr.19: 10869286-10869286 on Build GRCh38
rs756899297 Chr.19: 10869209-10869209 on Build GRCh38
rs548460163 Chr.19: 10869241-10869241 on Build GRCh38
rs988627362 Chr.19: 10869410-10869410 on Build GRCh38
rs944216673 Chr.19: 10869291-10869291 on Build GRCh38
rs767865908 Chr.19: 10869292-10869292 on Build GRCh38
rs1005869547 Chr.19: 10869377-10869377 on Build GRCh38
rs368631494 Chr.19: 10869296-10869296 on Build GRCh38
rs762189394 Chr.19: 10869240-10869240 on Build GRCh38
rs1032693636 Chr.19: 10869387-10869387 on Build GRCh38
rs887482926 Chr.19: 10869353-10869353 on Build GRCh38
rs779196018 Chr.19: 10869388-10869389 on Build GRCh38
rs955776332 Chr.19: 10869392-10869392 on Build GRCh38
rs777891207 Chr.19: 10869371-10869371 on Build GRCh38
rs749584957 Chr.19: 10869376-10869376 on Build GRCh38
rs537302791 Chr.19: 10869315-10869315 on Build GRCh38
rs754363930 Chr.19: 10869229-10869231 on Build GRCh38

View Full Product Details