Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 113619785-113620042 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAGI3
Entrez Gene ID
260425
Gene Name
membrane associated guanylate kinase, WW and PDZ domain containing 3
Gene Aliases
MAGI-3, dJ730K3.2
UniGene
Hs.486189
Gene Chromosome Location
Chr.1: 113390474-113685923 on Build GRCh38
Transcripts
NM_001142782.1 NM_152900.2 XM_005270737.3 XM_011541208.2 XM_017000974.1 XM_017000975.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2120186 c.1169C>T p.P390L Chr.1: 113619828-113619828 on Build GRCh38
2120187 c.1169C>T p.P390L Chr.1: 113619828-113619828 on Build GRCh38
6706345 c.1164delA p.K389fs*25 Chr.1: 113619823-113619823 on Build GRCh38
6706346 c.1164delA p.K389fs*25 Chr.1: 113619823-113619823 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs945307879 Chr.1: 113619983-113619983 on Build GRCh38
rs944515330 Chr.1: 113620001-113620001 on Build GRCh38
rs1038862951 Chr.1: 113620004-113620004 on Build GRCh38
rs113408933 Chr.1: 113619903-113619903 on Build GRCh38
rs752860412 Chr.1: 113619833-113619833 on Build GRCh38
rs539487902 Chr.1: 113619848-113619848 on Build GRCh38
rs913255670 Chr.1: 113619884-113619884 on Build GRCh38
rs370056115 Chr.1: 113619816-113619816 on Build GRCh38
rs764846621 Chr.1: 113619873-113619873 on Build GRCh38
rs927276989 Chr.1: 113619992-113619992 on Build GRCh38
rs112542389 Chr.1: 113619960-113619960 on Build GRCh38
rs781236048 Chr.1: 113619864-113619864 on Build GRCh38
rs990131698 Chr.1: 113619872-113619872 on Build GRCh38
rs758635350 Chr.1: 113619835-113619835 on Build GRCh38
rs1037817236 Chr.1: 113619972-113619972 on Build GRCh38
rs190724690 Chr.1: 113619943-113619943 on Build GRCh38
rs1217193 Chr.1: 113619997-113619997 on Build GRCh38
rs78351958 Chr.1: 113619929-113619929 on Build GRCh38
rs978005015 Chr.1: 113619985-113619985 on Build GRCh38
rs765591863 Chr.1: 113619828-113619828 on Build GRCh38
rs374433209 Chr.1: 113619847-113619847 on Build GRCh38

View Full Product Details